Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46782368-46782595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC1A5
Entrez Gene ID
6510
Gene Name
solute carrier family 1 member 5
Gene Aliases
AAAT, ASCT2, ATBO, M7V1, M7VS1, R16, RDRC
UniGene
Hs.631582
Gene Chromosome Location
Chr.19: 46759685-46788622 on Build GRCh38
Transcripts
NM_001145144.1 NM_001145145.1 NM_005628.2 XM_005259167.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780066 c.731C>T p.A244V Chr.19: 46782476-46782476 on Build GRCh38
4581245 c.717C>T p.I239I Chr.19: 46782490-46782490 on Build GRCh38
6314492 c.52-5C>A p.? Chr.19: 46782554-46782554 on Build GRCh38
4581246 c.111C>T p.I37I Chr.19: 46782490-46782490 on Build GRCh38
3536223 c.782C>T p.S261F Chr.19: 46782425-46782425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144011024 Chr.19: 46782490-46782490 on Build GRCh38
rs775276725 Chr.19: 46782419-46782419 on Build GRCh38
rs768599476 Chr.19: 46782543-46782543 on Build GRCh38
rs748831494 Chr.19: 46782476-46782476 on Build GRCh38
rs746119129 Chr.19: 46782564-46782564 on Build GRCh38
rs896748534 Chr.19: 46782556-46782556 on Build GRCh38
rs371513493 Chr.19: 46782466-46782466 on Build GRCh38
rs750535559 Chr.19: 46782441-46782441 on Build GRCh38
rs753332537 Chr.19: 46782571-46782571 on Build GRCh38
rs760563693 Chr.19: 46782541-46782541 on Build GRCh38
rs773215045 Chr.19: 46782544-46782544 on Build GRCh38
rs148681286 Chr.19: 46782438-46782438 on Build GRCh38
rs752162598 Chr.19: 46782426-46782426 on Build GRCh38
rs767413682 Chr.19: 46782410-46782410 on Build GRCh38
rs142211617 Chr.19: 46782449-46782449 on Build GRCh38
rs759263181 Chr.19: 46782409-46782409 on Build GRCh38
rs150805394 Chr.19: 46782450-46782450 on Build GRCh38
rs778533144 Chr.19: 46782573-46782573 on Build GRCh38
rs755297804 Chr.19: 46782463-46782463 on Build GRCh38
rs201808175 Chr.19: 46782566-46782566 on Build GRCh38
rs764933720 Chr.19: 46782405-46782405 on Build GRCh38
rs530038611 Chr.19: 46782455-46782455 on Build GRCh38
rs751720625 Chr.19: 46782552-46782552 on Build GRCh38
rs146442387 Chr.19: 46782508-46782508 on Build GRCh38
rs771740095 Chr.19: 46782492-46782492 on Build GRCh38
rs200463627 Chr.19: 46782437-46782437 on Build GRCh38
rs368539191 Chr.19: 46782397-46782397 on Build GRCh38
rs201988780 Chr.19: 46782428-46782428 on Build GRCh38
rs1005578254 Chr.19: 46782399-46782399 on Build GRCh38
rs375794946 Chr.19: 46782572-46782572 on Build GRCh38
rs775394315 Chr.19: 46782517-46782517 on Build GRCh38
rs770651202 Chr.19: 46782478-46782478 on Build GRCh38
rs753145096 Chr.19: 46782569-46782569 on Build GRCh38
rs759854931 Chr.19: 46782560-46782560 on Build GRCh38
rs943268758 Chr.19: 46782421-46782421 on Build GRCh38
rs140908383 Chr.19: 46782545-46782545 on Build GRCh38
rs748645826 Chr.19: 46782470-46782470 on Build GRCh38
rs139034381 Chr.19: 46782475-46782475 on Build GRCh38
rs956605349 Chr.19: 46782452-46782452 on Build GRCh38
rs757859365 Chr.19: 46782558-46782577 on Build GRCh38
rs774146143 Chr.19: 46782406-46782406 on Build GRCh38
rs778439337 Chr.19: 46782570-46782570 on Build GRCh38
rs756727391 Chr.19: 46782471-46782471 on Build GRCh38
rs200151640 Chr.19: 46782549-46782549 on Build GRCh38

View Full Product Details