Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154968081-154968306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHC1
Entrez Gene ID
6464
Gene Name
SHC adaptor protein 1
Gene Aliases
SHC, SHCA
UniGene
Hs.433795
Gene Chromosome Location
Chr.1: 154962298-154974492 on Build GRCh38
Transcripts
NM_001130040.1 NM_001130041.1 NM_001202859.1 NM_003029.4 NM_183001.4 XM_005245449.4 XM_005245451.4 XM_011509892.2 XM_011509893.2 XM_011509894.2 XM_011509897.1 XM_011509898.2 XM_017002081.1 XM_017002082.1 XM_017002083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7269007 c.426C>T p.I142I Chr.1: 154968252-154968252 on Build GRCh38
5296358 c.803C>T p.P268L Chr.1: 154968205-154968205 on Build GRCh38
7269006 c.756C>T p.I252I Chr.1: 154968252-154968252 on Build GRCh38
5296359 c.473C>T p.P158L Chr.1: 154968205-154968205 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs869145828 Chr.1: 154968182-154968182 on Build GRCh38
rs763850660 Chr.1: 154968210-154968210 on Build GRCh38
rs957896651 Chr.1: 154968119-154968119 on Build GRCh38
rs756658035 Chr.1: 154968273-154968275 on Build GRCh38
rs144359625 Chr.1: 154968219-154968219 on Build GRCh38
rs748359392 Chr.1: 154968162-154968162 on Build GRCh38
rs757577874 Chr.1: 154968135-154968135 on Build GRCh38
rs199885604 Chr.1: 154968203-154968203 on Build GRCh38
rs780741267 Chr.1: 154968237-154968237 on Build GRCh38
rs748470627 Chr.1: 154968270-154968270 on Build GRCh38
rs200764750 Chr.1: 154968179-154968179 on Build GRCh38
rs571607540 Chr.1: 154968131-154968131 on Build GRCh38
rs967585687 Chr.1: 154968174-154968174 on Build GRCh38
rs775365583 Chr.1: 154968188-154968188 on Build GRCh38
rs773162860 Chr.1: 154968178-154968178 on Build GRCh38
rs990397885 Chr.1: 154968123-154968123 on Build GRCh38
rs777016042 Chr.1: 154968155-154968155 on Build GRCh38
rs370145060 Chr.1: 154968185-154968185 on Build GRCh38
rs755443582 Chr.1: 154968249-154968249 on Build GRCh38
rs781668432 Chr.1: 154968266-154968266 on Build GRCh38
rs753078350 Chr.1: 154968174-154968176 on Build GRCh38
rs769959776 Chr.1: 154968166-154968166 on Build GRCh38
rs747472671 Chr.1: 154968243-154968243 on Build GRCh38
rs374354220 Chr.1: 154968279-154968279 on Build GRCh38
rs1020900747 Chr.1: 154968173-154968173 on Build GRCh38
rs983330421 Chr.1: 154968159-154968159 on Build GRCh38
rs143882709 Chr.1: 154968252-154968252 on Build GRCh38
rs757861390 Chr.1: 154968234-154968234 on Build GRCh38
rs753525154 Chr.1: 154968212-154968212 on Build GRCh38
rs939048697 Chr.1: 154968285-154968285 on Build GRCh38
rs756853675 Chr.1: 154968216-154968216 on Build GRCh38

View Full Product Details