Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49995048-49995319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VRK3
Entrez Gene ID
51231
Gene Name
vaccinia related kinase 3
Gene Aliases
-
UniGene
Hs.443330
Gene Chromosome Location
Chr.19: 49976467-50025954 on Build GRCh38
Transcripts
NM_001025778.1 NM_001308420.1 NM_016440.3 XM_005258971.3 XM_005258972.3 XM_006723237.3 XM_006723238.3 XM_011527023.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4080266 c.741C>T p.F247F Chr.19: 49995214-49995214 on Build GRCh38
3537085 c.748C>T p.H250Y Chr.19: 49995207-49995207 on Build GRCh38
4794549 c.736G>A p.G246S Chr.19: 49995219-49995219 on Build GRCh38
1318268 c.696G>A p.K232K Chr.19: 49995259-49995259 on Build GRCh38
5822795 c.733A>T p.M245L Chr.19: 49995222-49995222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776218659 Chr.19: 49995254-49995254 on Build GRCh38
rs1042110091 Chr.19: 49995112-49995112 on Build GRCh38
rs752024470 Chr.19: 49995195-49995195 on Build GRCh38
rs759566845 Chr.19: 49995171-49995171 on Build GRCh38
rs750722132 Chr.19: 49995181-49995181 on Build GRCh38
rs9304696 Chr.19: 49995071-49995071 on Build GRCh38
rs748321297 Chr.19: 49995214-49995214 on Build GRCh38
rs184543704 Chr.19: 49995251-49995251 on Build GRCh38
rs558019781 Chr.19: 49995235-49995235 on Build GRCh38
rs770798508 Chr.19: 49995230-49995230 on Build GRCh38
rs774475421 Chr.19: 49995162-49995162 on Build GRCh38
rs903303136 Chr.19: 49995123-49995123 on Build GRCh38
rs996732325 Chr.19: 49995182-49995182 on Build GRCh38
rs745731984 Chr.19: 49995241-49995241 on Build GRCh38
rs776470322 Chr.19: 49995103-49995103 on Build GRCh38
rs149272924 Chr.19: 49995159-49995159 on Build GRCh38
rs9304697 Chr.19: 49995131-49995131 on Build GRCh38
rs1003921852 Chr.19: 49995219-49995219 on Build GRCh38
rs766754308 Chr.19: 49995190-49995190 on Build GRCh38
rs772865245 Chr.19: 49995140-49995140 on Build GRCh38
rs768133471 Chr.19: 49995280-49995280 on Build GRCh38
rs768222370 Chr.19: 49995153-49995154 on Build GRCh38
rs1056031684 Chr.19: 49995090-49995090 on Build GRCh38
rs372977892 Chr.19: 49995183-49995183 on Build GRCh38
rs576877839 Chr.19: 49995110-49995110 on Build GRCh38
rs751937957 Chr.19: 49995276-49995276 on Build GRCh38
rs778559581 Chr.19: 49995282-49995282 on Build GRCh38
rs570016528 Chr.19: 49995250-49995250 on Build GRCh38
rs755483271 Chr.19: 49995197-49995197 on Build GRCh38
rs763873728 Chr.19: 49995175-49995175 on Build GRCh38
rs774848838 Chr.19: 49995247-49995247 on Build GRCh38
rs147531870 Chr.19: 49995243-49995243 on Build GRCh38
rs112433779 Chr.19: 49995160-49995160 on Build GRCh38
rs769910339 Chr.19: 49995274-49995274 on Build GRCh38
rs766666449 Chr.19: 49995271-49995271 on Build GRCh38
rs760187069 Chr.19: 49995174-49995174 on Build GRCh38
rs140220611 Chr.19: 49995256-49995256 on Build GRCh38
rs73055309 Chr.19: 49995083-49995083 on Build GRCh38
rs752685480 Chr.19: 49995281-49995281 on Build GRCh38
rs964697981 Chr.19: 49995109-49995109 on Build GRCh38
rs9304699 Chr.19: 49995298-49995298 on Build GRCh38
rs374964866 Chr.19: 49995229-49995229 on Build GRCh38
rs531327028 Chr.19: 49995094-49995094 on Build GRCh38
rs756263353 Chr.19: 49995293-49995293 on Build GRCh38
rs938368682 Chr.19: 49995186-49995186 on Build GRCh38
rs756282173 Chr.19: 49995228-49995228 on Build GRCh38
rs775069188 Chr.19: 49995173-49995173 on Build GRCh38
rs887937206 Chr.19: 49995074-49995074 on Build GRCh38
rs565911726 Chr.19: 49995213-49995213 on Build GRCh38
rs9304698 Chr.19: 49995295-49995295 on Build GRCh38
rs755467296 Chr.19: 49995279-49995279 on Build GRCh38
rs980033854 Chr.19: 49995176-49995176 on Build GRCh38

View Full Product Details