Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67408914-67409167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D10C
Entrez Gene ID
374403
Gene Name
TBC1 domain family member 10C
Gene Aliases
CARABIN, EPI64C
UniGene
Hs.534648
Gene Chromosome Location
Chr.11: 67403718-67410090 on Build GRCh38
Transcripts
NM_001256508.1 NM_198517.3 NR_046266.1 XM_006718538.3 XM_006718539.3 XM_006718540.3 XM_006718541.3 XM_006718542.3 XM_006718543.2 XM_006718544.3 XM_011545001.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613827 c.953_954insC p.A321fs*56 Chr.11: 67409093-67409094 on Build GRCh38
6443419 c.874G>A p.V292M Chr.11: 67409014-67409014 on Build GRCh38
5516575 c.947G>A p.R316Q Chr.11: 67409087-67409087 on Build GRCh38
5966122 c.873G>T p.L291L Chr.11: 67409013-67409013 on Build GRCh38
7393098 c.949G>C p.A317P Chr.11: 67409089-67409089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865854941 Chr.11: 67409077-67409077 on Build GRCh38
rs775695502 Chr.11: 67408997-67408997 on Build GRCh38
rs778937580 Chr.11: 67408937-67408937 on Build GRCh38
rs889506094 Chr.11: 67409045-67409045 on Build GRCh38
rs767141479 Chr.11: 67409024-67409024 on Build GRCh38
rs757340271 Chr.11: 67408964-67408964 on Build GRCh38
rs757401349 Chr.11: 67409093-67409094 on Build GRCh38
rs758395996 Chr.11: 67409074-67409074 on Build GRCh38
rs756209112 Chr.11: 67409137-67409137 on Build GRCh38
rs772927391 Chr.11: 67409025-67409025 on Build GRCh38
rs200282604 Chr.11: 67409113-67409113 on Build GRCh38
rs780355719 Chr.11: 67409089-67409089 on Build GRCh38
rs774946111 Chr.11: 67409017-67409017 on Build GRCh38
rs982110357 Chr.11: 67408938-67408938 on Build GRCh38
rs781233756 Chr.11: 67409081-67409081 on Build GRCh38
rs756296905 Chr.11: 67409087-67409087 on Build GRCh38
rs746085011 Chr.11: 67408995-67408995 on Build GRCh38
rs367546852 Chr.11: 67409018-67409018 on Build GRCh38
rs760393138 Chr.11: 67409026-67409026 on Build GRCh38
rs201172483 Chr.11: 67409014-67409014 on Build GRCh38
rs1034104580 Chr.11: 67409141-67409141 on Build GRCh38
rs989611661 Chr.11: 67409070-67409070 on Build GRCh38
rs372368867 Chr.11: 67409140-67409140 on Build GRCh38
rs888320898 Chr.11: 67408936-67408936 on Build GRCh38
rs753550059 Chr.11: 67409035-67409035 on Build GRCh38
rs368755663 Chr.11: 67409118-67409118 on Build GRCh38
rs371381997 Chr.11: 67409103-67409103 on Build GRCh38
rs959723860 Chr.11: 67408944-67408944 on Build GRCh38
rs781322580 Chr.11: 67408970-67408970 on Build GRCh38
rs745996977 Chr.11: 67409083-67409083 on Build GRCh38
rs558349429 Chr.11: 67408946-67408946 on Build GRCh38
rs765021942 Chr.11: 67409060-67409060 on Build GRCh38
rs375692810 Chr.11: 67409147-67409147 on Build GRCh38
rs976764282 Chr.11: 67409016-67409016 on Build GRCh38
rs770437810 Chr.11: 67409099-67409099 on Build GRCh38
rs140449515 Chr.11: 67409100-67409100 on Build GRCh38
rs1015545353 Chr.11: 67408947-67408947 on Build GRCh38
rs766187059 Chr.11: 67409028-67409028 on Build GRCh38
rs762054648 Chr.11: 67409013-67409013 on Build GRCh38
rs752545514 Chr.11: 67409102-67409102 on Build GRCh38
rs748664051 Chr.11: 67409097-67409097 on Build GRCh38
rs772830757 Chr.11: 67409098-67409098 on Build GRCh38
rs147560025 Chr.11: 67409094-67409094 on Build GRCh38
rs144309458 Chr.11: 67409101-67409101 on Build GRCh38
rs751687887 Chr.11: 67409094-67409094 on Build GRCh38
rs909718881 Chr.11: 67408969-67408969 on Build GRCh38
rs749566186 Chr.11: 67409093-67409093 on Build GRCh38
rs926723260 Chr.11: 67408940-67408940 on Build GRCh38
rs1027231344 Chr.11: 67408934-67408934 on Build GRCh38
rs868018194 Chr.11: 67409086-67409086 on Build GRCh38
rs754821458 Chr.11: 67409044-67409044 on Build GRCh38
rs576906702 Chr.11: 67408996-67408996 on Build GRCh38
rs554034570 Chr.11: 67409126-67409126 on Build GRCh38
rs752682113 Chr.11: 67409073-67409073 on Build GRCh38

View Full Product Details