Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176943100-176943373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UIMC1
Entrez Gene ID
51720
Gene Name
ubiquitin interaction motif containing 1
Gene Aliases
RAP80, X2HRIP110
UniGene
Hs.232721
Gene Chromosome Location
Chr.5: 176905005-177022640 on Build GRCh38
Transcripts
NM_001199297.2 NM_001199298.1 NM_001317961.1 NM_016290.4 XM_005265930.2 XM_005265932.2 XM_005265933.2 XM_005265934.2 XM_005265935.2 XM_005265936.2 XM_006714871.2 XM_006714874.2 XM_011534568.2 XM_011534570.1 XM_017009574.1 XM_017009575.1 XM_017009576.1 XM_017009577.1 XM_017009578.1 XM_017009579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394399 c.1576G>T p.G526C Chr.5: 176943356-176943356 on Build GRCh38
6170534 c.1561G>A p.A521T Chr.5: 176943371-176943371 on Build GRCh38
6300336 c.1560T>C p.H520H Chr.5: 176943372-176943372 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545077013 Chr.5: 176943341-176943341 on Build GRCh38
rs998488118 Chr.5: 176943154-176943154 on Build GRCh38
rs781240127 Chr.5: 176943347-176943347 on Build GRCh38
rs200945557 Chr.5: 176943348-176943348 on Build GRCh38
rs776872461 Chr.5: 176943306-176943307 on Build GRCh38
rs1033997281 Chr.5: 176943143-176943143 on Build GRCh38
rs951555463 Chr.5: 176943255-176943255 on Build GRCh38
rs555579908 Chr.5: 176943129-176943129 on Build GRCh38
rs757567980 Chr.5: 176943306-176943306 on Build GRCh38
rs754230914 Chr.5: 176943297-176943297 on Build GRCh38
rs956530248 Chr.5: 176943265-176943265 on Build GRCh38
rs755666630 Chr.5: 176943339-176943339 on Build GRCh38
rs553209700 Chr.5: 176943261-176943261 on Build GRCh38
rs1017089270 Chr.5: 176943320-176943320 on Build GRCh38
rs764517294 Chr.5: 176943296-176943296 on Build GRCh38
rs372555749 Chr.5: 176943311-176943311 on Build GRCh38
rs752704018 Chr.5: 176943281-176943281 on Build GRCh38
rs757983875 Chr.5: 176943329-176943329 on Build GRCh38
rs894180967 Chr.5: 176943177-176943177 on Build GRCh38
rs962752007 Chr.5: 176943326-176943326 on Build GRCh38
rs1024980684 Chr.5: 176943246-176943246 on Build GRCh38
rs578026512 Chr.5: 176943298-176943298 on Build GRCh38
rs1018323788 Chr.5: 176943125-176943125 on Build GRCh38
rs761255160 Chr.5: 176943294-176943294 on Build GRCh38
rs149170785 Chr.5: 176943139-176943139 on Build GRCh38
rs1006566666 Chr.5: 176943124-176943124 on Build GRCh38
rs746449782 Chr.5: 176943340-176943340 on Build GRCh38
rs533450616 Chr.5: 176943211-176943211 on Build GRCh38
rs369156033 Chr.5: 176943332-176943332 on Build GRCh38
rs535155375 Chr.5: 176943202-176943202 on Build GRCh38
rs779674163 Chr.5: 176943330-176943330 on Build GRCh38
rs376298872 Chr.5: 176943315-176943315 on Build GRCh38
rs771260769 Chr.5: 176943297-176943297 on Build GRCh38

View Full Product Details