Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52230797-52231022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TWF2
Entrez Gene ID
11344
Gene Name
twinfilin actin binding protein 2
Gene Aliases
A6RP, A6r, MSTP011, PTK9L
UniGene
Hs.436439
Gene Chromosome Location
Chr.3: 52228610-52239167 on Build GRCh38
Transcripts
NM_007284.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186311 c.535G>A p.A179T Chr.3: 52230944-52230944 on Build GRCh38
3760162 c.588A>G p.K196K Chr.3: 52230891-52230891 on Build GRCh38
3595897 c.570C>T p.L190L Chr.3: 52230909-52230909 on Build GRCh38
3595896 c.604C>T p.Q202* Chr.3: 52230875-52230875 on Build GRCh38
5399708 c.540C>T p.F180F Chr.3: 52230939-52230939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114935680 Chr.3: 52230943-52230943 on Build GRCh38
rs773240482 Chr.3: 52230885-52230885 on Build GRCh38
rs777047645 Chr.3: 52230907-52230907 on Build GRCh38
rs778651117 Chr.3: 52230968-52230968 on Build GRCh38
rs765720266 Chr.3: 52230917-52230917 on Build GRCh38
rs748125935 Chr.3: 52230855-52230855 on Build GRCh38
rs766715944 Chr.3: 52230903-52230903 on Build GRCh38
rs957641164 Chr.3: 52230954-52230954 on Build GRCh38
rs352143 Chr.3: 52230891-52230891 on Build GRCh38
rs755910062 Chr.3: 52230833-52230833 on Build GRCh38
rs774464530 Chr.3: 52230844-52230844 on Build GRCh38
rs780012928 Chr.3: 52230839-52230839 on Build GRCh38
rs187719121 Chr.3: 52230960-52230960 on Build GRCh38
rs551290450 Chr.3: 52230981-52230981 on Build GRCh38
rs373959049 Chr.3: 52230985-52230985 on Build GRCh38
rs779994594 Chr.3: 52230945-52230945 on Build GRCh38
rs761159375 Chr.3: 52230919-52230919 on Build GRCh38
rs753746677 Chr.3: 52230951-52230951 on Build GRCh38
rs758622058 Chr.3: 52230827-52230827 on Build GRCh38
rs771997419 Chr.3: 52230976-52230976 on Build GRCh38
rs748149583 Chr.3: 52230971-52230971 on Build GRCh38
rs767082492 Chr.3: 52230931-52230931 on Build GRCh38
rs201759490 Chr.3: 52230916-52230916 on Build GRCh38
rs747201043 Chr.3: 52231003-52231003 on Build GRCh38
rs184548327 Chr.3: 52230842-52230842 on Build GRCh38
rs375895610 Chr.3: 52230944-52230944 on Build GRCh38
rs924377390 Chr.3: 52230828-52230828 on Build GRCh38
rs375378648 Chr.3: 52230843-52230843 on Build GRCh38
rs375522201 Chr.3: 52230861-52230861 on Build GRCh38
rs898704194 Chr.3: 52230989-52230989 on Build GRCh38
rs964799810 Chr.3: 52230866-52230866 on Build GRCh38

View Full Product Details