Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10404577-10404844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFAP2A
Entrez Gene ID
7020
Gene Name
transcription factor AP-2 alpha
Gene Aliases
AP-2, AP-2alpha, AP2TF, BOFS, TFAP2
UniGene
Hs.519880
Gene Chromosome Location
Chr.6: 10396677-10419897 on Build GRCh38
Transcripts
NM_001032280.2 NM_001042425.1 NM_003220.2 XM_006715175.2 XM_011514833.2 XM_017011232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996579 c.638C>T p.S213L Chr.6: 10404634-10404634 on Build GRCh38
6800336 c.612C>T p.N204N Chr.6: 10404648-10404648 on Build GRCh38
6105004 c.554C>G p.S185C Chr.6: 10404700-10404700 on Build GRCh38
1672231 c.646G>T p.G216C Chr.6: 10404626-10404626 on Build GRCh38
5595429 c.611A>G p.N204S Chr.6: 10404649-10404649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs793888541 Chr.6: 10404631-10404631 on Build GRCh38
rs1041684112 Chr.6: 10404726-10404726 on Build GRCh38
rs777922831 Chr.6: 10404642-10404642 on Build GRCh38
rs138809786 Chr.6: 10404793-10404793 on Build GRCh38
rs370522296 Chr.6: 10404777-10404777 on Build GRCh38
rs746862188 Chr.6: 10404672-10404672 on Build GRCh38
rs372587710 Chr.6: 10404681-10404681 on Build GRCh38
rs531975577 Chr.6: 10404735-10404735 on Build GRCh38
rs768098328 Chr.6: 10404599-10404599 on Build GRCh38
rs1019630699 Chr.6: 10404821-10404821 on Build GRCh38
rs979214687 Chr.6: 10404634-10404634 on Build GRCh38
rs1052818666 Chr.6: 10404720-10404720 on Build GRCh38
rs375564449 Chr.6: 10404758-10404758 on Build GRCh38
rs548526516 Chr.6: 10404813-10404813 on Build GRCh38
rs751970882 Chr.6: 10404753-10404753 on Build GRCh38
rs761543465 Chr.6: 10404687-10404687 on Build GRCh38
rs146735451 Chr.6: 10404724-10404724 on Build GRCh38
rs772033707 Chr.6: 10404669-10404669 on Build GRCh38
rs768437372 Chr.6: 10404677-10404677 on Build GRCh38
rs776516420 Chr.6: 10404788-10404788 on Build GRCh38
rs376887906 Chr.6: 10404762-10404762 on Build GRCh38
rs529866454 Chr.6: 10404810-10404810 on Build GRCh38
rs767853436 Chr.6: 10404801-10404812 on Build GRCh38
rs745892262 Chr.6: 10404666-10404666 on Build GRCh38
rs780030798 Chr.6: 10404774-10404774 on Build GRCh38
rs909713555 Chr.6: 10404637-10404637 on Build GRCh38
rs1033181674 Chr.6: 10404793-10404793 on Build GRCh38
rs925135582 Chr.6: 10404684-10404684 on Build GRCh38
rs755569103 Chr.6: 10404597-10404597 on Build GRCh38
rs201296512 Chr.6: 10404716-10404716 on Build GRCh38
rs772810778 Chr.6: 10404695-10404695 on Build GRCh38
rs905925169 Chr.6: 10404787-10404787 on Build GRCh38
rs541264751 Chr.6: 10404748-10404748 on Build GRCh38
rs771955798 Chr.6: 10404715-10404720 on Build GRCh38
rs756526526 Chr.6: 10404621-10404621 on Build GRCh38
rs757530487 Chr.6: 10404742-10404742 on Build GRCh38
rs563554662 Chr.6: 10404814-10404814 on Build GRCh38
rs793888540 Chr.6: 10404623-10404623 on Build GRCh38
rs369302647 Chr.6: 10404618-10404618 on Build GRCh38
rs749528750 Chr.6: 10404644-10404644 on Build GRCh38
rs887977416 Chr.6: 10404804-10404805 on Build GRCh38
rs758457244 Chr.6: 10404773-10404773 on Build GRCh38
rs763437712 Chr.6: 10404664-10404664 on Build GRCh38
rs764532449 Chr.6: 10404727-10404727 on Build GRCh38
rs768525762 Chr.6: 10404784-10404784 on Build GRCh38
rs143363077 Chr.6: 10404699-10404699 on Build GRCh38
rs764751482 Chr.6: 10404692-10404692 on Build GRCh38
rs565071190 Chr.6: 10404709-10404709 on Build GRCh38
rs903307290 Chr.6: 10404743-10404743 on Build GRCh38
rs966300825 Chr.6: 10404825-10404825 on Build GRCh38
rs953677570 Chr.6: 10404803-10404803 on Build GRCh38
rs202002859 Chr.6: 10404649-10404649 on Build GRCh38

View Full Product Details