Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5967253-5967524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWF
Entrez Gene ID
7450
Gene Name
von Willebrand factor
Gene Aliases
F8VWF, VWD
UniGene
Hs.440848
Gene Chromosome Location
Chr.12: 5948874-6124675 on Build GRCh38
Transcripts
NM_000552.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5968631 c.7883C>A p.P2628H Chr.12: 5967490-5967490 on Build GRCh38
6879810 c.7883C>T p.P2628L Chr.12: 5967490-5967490 on Build GRCh38
5040310 c.7865C>T p.T2622I Chr.12: 5967508-5967508 on Build GRCh38
3464058 c.7860G>A p.R2620R Chr.12: 5967513-5967513 on Build GRCh38
1493000 c.7886T>C p.L2629P Chr.12: 5967487-5967487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748622872 Chr.12: 5967489-5967489 on Build GRCh38
rs113814258 Chr.12: 5967484-5967484 on Build GRCh38
rs769041752 Chr.12: 5967330-5967330 on Build GRCh38
rs1042172767 Chr.12: 5967359-5967359 on Build GRCh38
rs751768351 Chr.12: 5967445-5967445 on Build GRCh38
rs547720280 Chr.12: 5967388-5967388 on Build GRCh38
rs748257902 Chr.12: 5967366-5967366 on Build GRCh38
rs141666705 Chr.12: 5967497-5967497 on Build GRCh38
rs552486045 Chr.12: 5967369-5967369 on Build GRCh38
rs1024570750 Chr.12: 5967333-5967333 on Build GRCh38
rs982182587 Chr.12: 5967380-5967380 on Build GRCh38
rs926333904 Chr.12: 5967394-5967397 on Build GRCh38
rs762917566 Chr.12: 5967443-5967443 on Build GRCh38
rs1027354339 Chr.12: 5967491-5967491 on Build GRCh38
rs1010452956 Chr.12: 5967326-5967326 on Build GRCh38
rs770381255 Chr.12: 5967492-5967492 on Build GRCh38
rs565651433 Chr.12: 5967389-5967390 on Build GRCh38
rs201118818 Chr.12: 5967463-5967463 on Build GRCh38
rs532478457 Chr.12: 5967328-5967328 on Build GRCh38
rs773560179 Chr.12: 5967494-5967494 on Build GRCh38
rs370959569 Chr.12: 5967444-5967444 on Build GRCh38
rs374631669 Chr.12: 5967446-5967446 on Build GRCh38
rs533941161 Chr.12: 5967374-5967374 on Build GRCh38
rs752608230 Chr.12: 5967455-5967455 on Build GRCh38
rs756033498 Chr.12: 5967460-5967460 on Build GRCh38
rs185085412 Chr.12: 5967412-5967412 on Build GRCh38
rs778762511 Chr.12: 5967471-5967471 on Build GRCh38
rs200573272 Chr.12: 5967462-5967462 on Build GRCh38
rs112100565 Chr.12: 5967487-5967487 on Build GRCh38
rs145180946 Chr.12: 5967311-5967311 on Build GRCh38
rs571713194 Chr.12: 5967370-5967370 on Build GRCh38
rs745458385 Chr.12: 5967473-5967473 on Build GRCh38
rs902372127 Chr.12: 5967363-5967363 on Build GRCh38
rs55687637 Chr.12: 5967474-5967474 on Build GRCh38
rs781035747 Chr.12: 5967354-5967354 on Build GRCh38

View Full Product Details