Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108260809-108261054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX3
Entrez Gene ID
8724
Gene Name
sorting nexin 3
Gene Aliases
Grd19, MCOPS8, SDP3
UniGene
Hs.12102
Gene Chromosome Location
Chr.6: 108211217-108261260 on Build GRCh38
Transcripts
NM_001300928.1 NM_001300929.1 NM_003795.5 NM_152827.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6786585 c.75delC p.S26fs*8 Chr.6: 108260847-108260847 on Build GRCh38
6360032 c.14T>A p.V5E Chr.6: 108260908-108260908 on Build GRCh38
6360033 c.14T>A p.V5E Chr.6: 108260908-108260908 on Build GRCh38
6247913 c.5C>A p.A2E Chr.6: 108260917-108260917 on Build GRCh38
6247912 c.5C>A p.A2E Chr.6: 108260917-108260917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746047857 Chr.6: 108260969-108260970 on Build GRCh38
rs751463571 Chr.6: 108260936-108260936 on Build GRCh38
rs1003719362 Chr.6: 108260847-108260847 on Build GRCh38
rs758398711 Chr.6: 108260943-108260944 on Build GRCh38
rs772523736 Chr.6: 108260895-108260895 on Build GRCh38
rs758212848 Chr.6: 108260855-108260855 on Build GRCh38
rs11537587 Chr.6: 108260902-108260902 on Build GRCh38
rs766349492 Chr.6: 108260934-108260934 on Build GRCh38
rs752644620 Chr.6: 108260943-108260943 on Build GRCh38
rs1031186679 Chr.6: 108260975-108260983 on Build GRCh38
rs907775123 Chr.6: 108261022-108261022 on Build GRCh38
rs769387256 Chr.6: 108260962-108260962 on Build GRCh38
rs765982967 Chr.6: 108260933-108260934 on Build GRCh38
rs766332606 Chr.6: 108260859-108260859 on Build GRCh38
rs777761818 Chr.6: 108260944-108260965 on Build GRCh38
rs11537584 Chr.6: 108260910-108260910 on Build GRCh38
rs563123307 Chr.6: 108260999-108260999 on Build GRCh38
rs780747111 Chr.6: 108260950-108260950 on Build GRCh38
rs753489451 Chr.6: 108260936-108260937 on Build GRCh38
rs774682097 Chr.6: 108260930-108260931 on Build GRCh38
rs764111559 Chr.6: 108260938-108260938 on Build GRCh38
rs373380139 Chr.6: 108260944-108260944 on Build GRCh38
rs759410964 Chr.6: 108260972-108260972 on Build GRCh38
rs761712839 Chr.6: 108260836-108260836 on Build GRCh38
rs142951692 Chr.6: 108260970-108260975 on Build GRCh38
rs756160135 Chr.6: 108260873-108260873 on Build GRCh38
rs747482657 Chr.6: 108260951-108260951 on Build GRCh38
rs754467261 Chr.6: 108260937-108260939 on Build GRCh38
rs778006414 Chr.6: 108260877-108260877 on Build GRCh38
rs772818252 Chr.6: 108260930-108260930 on Build GRCh38
rs750399984 Chr.6: 108260850-108260850 on Build GRCh38
rs746389539 Chr.6: 108260892-108260892 on Build GRCh38
rs943478744 Chr.6: 108261014-108261014 on Build GRCh38
rs779192511 Chr.6: 108260942-108260942 on Build GRCh38
rs772896682 Chr.6: 108260965-108260965 on Build GRCh38
rs749790700 Chr.6: 108261005-108261005 on Build GRCh38
rs796663783 Chr.6: 108260974-108260975 on Build GRCh38
rs761327116 Chr.6: 108260927-108260927 on Build GRCh38
rs920627925 Chr.6: 108261019-108261019 on Build GRCh38
rs948171192 Chr.6: 108260967-108260967 on Build GRCh38
rs573927562 Chr.6: 108261002-108261002 on Build GRCh38
rs760258383 Chr.6: 108260829-108260829 on Build GRCh38
rs780695887 Chr.6: 108260901-108260901 on Build GRCh38
rs1031589769 Chr.6: 108260979-108260979 on Build GRCh38
rs757079966 Chr.6: 108260950-108260951 on Build GRCh38
rs754018575 Chr.6: 108260886-108260886 on Build GRCh38
rs762692219 Chr.6: 108260968-108260968 on Build GRCh38
rs987333856 Chr.6: 108260987-108260987 on Build GRCh38
rs762793625 Chr.6: 108260932-108260932 on Build GRCh38
rs776400846 Chr.6: 108260835-108260835 on Build GRCh38
rs754102870 Chr.6: 108260939-108260939 on Build GRCh38
rs557400461 Chr.6: 108260959-108260959 on Build GRCh38
rs747844488 Chr.6: 108260961-108260961 on Build GRCh38
rs776458119 Chr.6: 108260926-108260926 on Build GRCh38
rs968493134 Chr.6: 108260990-108260992 on Build GRCh38
rs768249925 Chr.6: 108260915-108260915 on Build GRCh38
rs763898445 Chr.6: 108260832-108260832 on Build GRCh38
rs907806563 Chr.6: 108260921-108260921 on Build GRCh38
rs760772067 Chr.6: 108260937-108260937 on Build GRCh38
rs540671479 Chr.6: 108260974-108260975 on Build GRCh38
rs1011059133 Chr.6: 108260985-108260985 on Build GRCh38
rs911712507 Chr.6: 108261013-108261013 on Build GRCh38
rs369856168 Chr.6: 108260928-108260928 on Build GRCh38
rs758768172 Chr.6: 108260945-108260945 on Build GRCh38
rs577429439 Chr.6: 108260980-108260980 on Build GRCh38
rs752829628 Chr.6: 108260862-108260862 on Build GRCh38
rs770668528 Chr.6: 108260971-108260971 on Build GRCh38
rs974794941 Chr.6: 108261015-108261015 on Build GRCh38
rs764904684 Chr.6: 108260844-108260844 on Build GRCh38
rs1028752956 Chr.6: 108261012-108261012 on Build GRCh38
rs746773208 Chr.6: 108260949-108260950 on Build GRCh38
rs781591422 Chr.6: 108260952-108260953 on Build GRCh38
rs996288516 Chr.6: 108261000-108261000 on Build GRCh38
rs543246460 Chr.6: 108260983-108260983 on Build GRCh38
rs962298684 Chr.6: 108261024-108261024 on Build GRCh38
rs1022917915 Chr.6: 108260989-108260989 on Build GRCh38
rs534716393 Chr.6: 108260940-108260940 on Build GRCh38
rs780794151 Chr.6: 108260960-108260960 on Build GRCh38
rs779088331 Chr.6: 108260889-108260889 on Build GRCh38

View Full Product Details