Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76013673-76013946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STYXL1
Entrez Gene ID
51657
Gene Name
serine/threonine/tyrosine interacting-like 1
Gene Aliases
DUSP24, MK-STYX, MKSTYX
UniGene
Hs.11615
Gene Chromosome Location
Chr.7: 75995820-76048709 on Build GRCh38
Transcripts
NM_001317785.1 NM_001317786.1 NM_001317787.1 NM_001317788.1 NM_001317789.1 NM_016086.2 NR_134486.1 NR_134487.1 XM_011516292.2 XM_011516293.2 XM_011516300.1 XM_017012296.1 XM_017012297.1 XM_017012298.1 XM_017012299.1 XM_017012300.1 XM_017012301.1 XM_017012302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434750 c.379C>A p.L127M Chr.7: 76013816-76013816 on Build GRCh38
3995702 c.308A>G p.D103G Chr.7: 76013887-76013887 on Build GRCh38
6792770 c.391T>A p.Y131N Chr.7: 76013804-76013804 on Build GRCh38
3882442 c.392A>G p.Y131C Chr.7: 76013803-76013803 on Build GRCh38
4903507 c.438C>T p.I146I Chr.7: 76013757-76013757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782158715 Chr.7: 76013718-76013718 on Build GRCh38
rs782741343 Chr.7: 76013736-76013736 on Build GRCh38
rs782122922 Chr.7: 76013826-76013826 on Build GRCh38
rs782434827 Chr.7: 76013770-76013770 on Build GRCh38
rs782533406 Chr.7: 76013852-76013852 on Build GRCh38
rs781930504 Chr.7: 76013732-76013732 on Build GRCh38
rs782700697 Chr.7: 76013728-76013728 on Build GRCh38
rs367788597 Chr.7: 76013879-76013879 on Build GRCh38
rs781869678 Chr.7: 76013846-76013846 on Build GRCh38
rs782281048 Chr.7: 76013788-76013788 on Build GRCh38
rs375103901 Chr.7: 76013916-76013916 on Build GRCh38
rs199592284 Chr.7: 76013707-76013707 on Build GRCh38
rs782334223 Chr.7: 76013819-76013819 on Build GRCh38
rs1004104030 Chr.7: 76013724-76013724 on Build GRCh38
rs368662024 Chr.7: 76013894-76013894 on Build GRCh38
rs782185648 Chr.7: 76013811-76013811 on Build GRCh38
rs373949822 Chr.7: 76013771-76013771 on Build GRCh38
rs782675673 Chr.7: 76013701-76013701 on Build GRCh38
rs781912400 Chr.7: 76013799-76013799 on Build GRCh38
rs756860207 Chr.7: 76013889-76013889 on Build GRCh38
rs782602977 Chr.7: 76013893-76013893 on Build GRCh38
rs782017244 Chr.7: 76013716-76013716 on Build GRCh38
rs782794725 Chr.7: 76013760-76013760 on Build GRCh38
rs781909480 Chr.7: 76013762-76013762 on Build GRCh38
rs201838555 Chr.7: 76013833-76013833 on Build GRCh38
rs376957588 Chr.7: 76013695-76013695 on Build GRCh38
rs782381495 Chr.7: 76013713-76013713 on Build GRCh38
rs782700056 Chr.7: 76013723-76013723 on Build GRCh38
rs781864399 Chr.7: 76013743-76013743 on Build GRCh38
rs186469256 Chr.7: 76013710-76013710 on Build GRCh38
rs372636375 Chr.7: 76013911-76013911 on Build GRCh38
rs782374162 Chr.7: 76013899-76013899 on Build GRCh38
rs781959500 Chr.7: 76013825-76013825 on Build GRCh38
rs782708721 Chr.7: 76013841-76013841 on Build GRCh38
rs374193256 Chr.7: 76013860-76013860 on Build GRCh38
rs370786510 Chr.7: 76013827-76013827 on Build GRCh38
rs782425576 Chr.7: 76013920-76013920 on Build GRCh38
rs367636888 Chr.7: 76013921-76013921 on Build GRCh38
rs782024429 Chr.7: 76013828-76013828 on Build GRCh38
rs782347485 Chr.7: 76013803-76013803 on Build GRCh38
rs368425776 Chr.7: 76013798-76013798 on Build GRCh38
rs782748308 Chr.7: 76013843-76013843 on Build GRCh38
rs782175555 Chr.7: 76013875-76013875 on Build GRCh38
rs782646288 Chr.7: 76013785-76013785 on Build GRCh38
rs782079568 Chr.7: 76013735-76013735 on Build GRCh38
rs782503460 Chr.7: 76013793-76013794 on Build GRCh38
rs537138292 Chr.7: 76013891-76013891 on Build GRCh38
rs782534280 Chr.7: 76013745-76013745 on Build GRCh38

View Full Product Details