Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52554433-52554660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF808
Entrez Gene ID
388558
Gene Name
zinc finger protein 808
Gene Aliases
-
UniGene
Hs.729294
Gene Chromosome Location
Chr.19: 52527656-52566018 on Build GRCh38
Transcripts
NM_001039886.3 NM_001321424.1 NM_001321425.1 XM_005258909.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1000590 c.1651A>G p.N551D Chr.19: 52554567-52554567 on Build GRCh38
1395965 c.1723G>A p.A575T Chr.19: 52554639-52554639 on Build GRCh38
1000589 c.1600A>G p.T534A Chr.19: 52554516-52554516 on Build GRCh38
1395964 c.1720A>T p.K574* Chr.19: 52554636-52554636 on Build GRCh38
4745054 c.1606G>T p.E536* Chr.19: 52554522-52554522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201823180 Chr.19: 52554465-52554465 on Build GRCh38
rs773473871 Chr.19: 52554519-52554519 on Build GRCh38
rs775859164 Chr.19: 52554493-52554493 on Build GRCh38
rs144870963 Chr.19: 52554469-52554469 on Build GRCh38
rs144248619 Chr.19: 52554516-52554516 on Build GRCh38
rs759739610 Chr.19: 52554524-52554525 on Build GRCh38
rs1019328632 Chr.19: 52554616-52554616 on Build GRCh38
rs755813973 Chr.19: 52554560-52554560 on Build GRCh38
rs766397222 Chr.19: 52554463-52554463 on Build GRCh38
rs748498322 Chr.19: 52554617-52554617 on Build GRCh38
rs766490209 Chr.19: 52554533-52554533 on Build GRCh38
rs868229788 Chr.19: 52554491-52554491 on Build GRCh38
rs376417824 Chr.19: 52554505-52554505 on Build GRCh38
rs549096847 Chr.19: 52554477-52554477 on Build GRCh38
rs754724436 Chr.19: 52554466-52554466 on Build GRCh38
rs780627649 Chr.19: 52554565-52554565 on Build GRCh38
rs776944248 Chr.19: 52554517-52554518 on Build GRCh38
rs777394484 Chr.19: 52554561-52554561 on Build GRCh38
rs745417432 Chr.19: 52554572-52554572 on Build GRCh38
rs769094182 Chr.19: 52554578-52554578 on Build GRCh38
rs147791268 Chr.19: 52554627-52554627 on Build GRCh38
rs141948878 Chr.19: 52554483-52554483 on Build GRCh38
rs199579964 Chr.19: 52554602-52554602 on Build GRCh38
rs867884275 Chr.19: 52554614-52554615 on Build GRCh38
rs778383397 Chr.19: 52554554-52554554 on Build GRCh38
rs867250965 Chr.19: 52554507-52554507 on Build GRCh38
rs753416971 Chr.19: 52554563-52554563 on Build GRCh38
rs185737879 Chr.19: 52554541-52554541 on Build GRCh38
rs759399598 Chr.19: 52554546-52554546 on Build GRCh38
rs199977771 Chr.19: 52554557-52554557 on Build GRCh38
rs375208140 Chr.19: 52554478-52554478 on Build GRCh38
rs777305164 Chr.19: 52554481-52554481 on Build GRCh38
rs757887789 Chr.19: 52554609-52554609 on Build GRCh38
rs773563625 Chr.19: 52554632-52554632 on Build GRCh38
rs772629320 Chr.19: 52554484-52554484 on Build GRCh38
rs192222088 Chr.19: 52554498-52554498 on Build GRCh38
rs373375731 Chr.19: 52554564-52554564 on Build GRCh38
rs537179814 Chr.19: 52554569-52554569 on Build GRCh38
rs756952792 Chr.19: 52554524-52554524 on Build GRCh38
rs765329038 Chr.19: 52554615-52554616 on Build GRCh38
rs747407810 Chr.19: 52554495-52554495 on Build GRCh38
rs771307577 Chr.19: 52554461-52554462 on Build GRCh38
rs550545156 Chr.19: 52554542-52554542 on Build GRCh38
rs770105641 Chr.19: 52554515-52554515 on Build GRCh38
rs776930729 Chr.19: 52554504-52554504 on Build GRCh38
rs753213177 Chr.19: 52554616-52554617 on Build GRCh38

View Full Product Details