Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 1072103-1072340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR37
Entrez Gene ID
22884
Gene Name
WD repeat domain 37
Gene Aliases
-
UniGene
Hs.188495
Gene Chromosome Location
Chr.10: 1056836-1132297 on Build GRCh38
Transcripts
NM_014023.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388140 c.76C>T p.R26* Chr.10: 1072231-1072231 on Build GRCh38
1270394 c.43A>C p.K15Q Chr.10: 1072198-1072198 on Build GRCh38
4011538 c.60C>A p.S20S Chr.10: 1072215-1072215 on Build GRCh38
427066 c.107C>T p.T36M Chr.10: 1072262-1072262 on Build GRCh38
6821800 c.67C>A p.L23I Chr.10: 1072222-1072222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566670086 Chr.10: 1072300-1072300 on Build GRCh38
rs373937534 Chr.10: 1072182-1072182 on Build GRCh38
rs778442799 Chr.10: 1072190-1072190 on Build GRCh38
rs556067345 Chr.10: 1072168-1072168 on Build GRCh38
rs759250769 Chr.10: 1072222-1072222 on Build GRCh38
rs375901168 Chr.10: 1072247-1072247 on Build GRCh38
rs752384383 Chr.10: 1072305-1072305 on Build GRCh38
rs751039874 Chr.10: 1072170-1072170 on Build GRCh38
rs762124997 Chr.10: 1072246-1072246 on Build GRCh38
rs774499095 Chr.10: 1072231-1072231 on Build GRCh38
rs200657141 Chr.10: 1072181-1072181 on Build GRCh38
rs770874601 Chr.10: 1072211-1072211 on Build GRCh38
rs772676409 Chr.10: 1072205-1072205 on Build GRCh38
rs776266116 Chr.10: 1072133-1072133 on Build GRCh38
rs769578695 Chr.10: 1072228-1072228 on Build GRCh38
rs760789787 Chr.10: 1072259-1072259 on Build GRCh38
rs771635221 Chr.10: 1072202-1072202 on Build GRCh38
rs992068746 Chr.10: 1072183-1072183 on Build GRCh38
rs376115508 Chr.10: 1072138-1072138 on Build GRCh38
rs199854657 Chr.10: 1072208-1072208 on Build GRCh38
rs757430730 Chr.10: 1072163-1072163 on Build GRCh38
rs747822059 Chr.10: 1072196-1072196 on Build GRCh38
rs767695186 Chr.10: 1072169-1072169 on Build GRCh38
rs755329682 Chr.10: 1072189-1072189 on Build GRCh38
rs751734034 Chr.10: 1072161-1072161 on Build GRCh38
rs370793268 Chr.10: 1072171-1072171 on Build GRCh38
rs201002615 Chr.10: 1072263-1072263 on Build GRCh38
rs764251023 Chr.10: 1072144-1072144 on Build GRCh38
rs17856557 Chr.10: 1072186-1072186 on Build GRCh38
rs775403198 Chr.10: 1072262-1072262 on Build GRCh38
rs916046990 Chr.10: 1072194-1072194 on Build GRCh38
rs750509895 Chr.10: 1072250-1072250 on Build GRCh38
rs947829094 Chr.10: 1072207-1072207 on Build GRCh38
rs370872470 Chr.10: 1072279-1072279 on Build GRCh38
rs776588127 Chr.10: 1072218-1072218 on Build GRCh38

View Full Product Details