Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160619675-160619948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLAMF1
Entrez Gene ID
6504
Gene Name
signaling lymphocytic activation molecule family member 1
Gene Aliases
CD150, CDw150, SLAM
UniGene
Hs.523660
Gene Chromosome Location
Chr.1: 160608103-160647537 on Build GRCh38
Transcripts
NM_003037.3 NR_104399.1 NR_104400.1 NR_104401.1 XM_005245456.3 XM_011509905.2 XM_017002129.1 XM_017002130.1 XM_017002131.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
126189 c.844G>A p.A282T Chr.1: 160619796-160619796 on Build GRCh38
2085986 c.798G>A p.T266T Chr.1: 160619842-160619842 on Build GRCh38
1335531 c.829delA p.S277fs*54 Chr.1: 160619811-160619811 on Build GRCh38
6779148 c.791-2A>G p.? Chr.1: 160619851-160619851 on Build GRCh38
382981 c.819G>A p.V273V Chr.1: 160619821-160619821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201439748 Chr.1: 160619819-160619819 on Build GRCh38
rs773246801 Chr.1: 160619842-160619842 on Build GRCh38
rs6693001 Chr.1: 160619892-160619892 on Build GRCh38
rs1021635763 Chr.1: 160619781-160619781 on Build GRCh38
rs759595270 Chr.1: 160619858-160619858 on Build GRCh38
rs747581992 Chr.1: 160619803-160619803 on Build GRCh38
rs767513275 Chr.1: 160619872-160619872 on Build GRCh38
rs755511002 Chr.1: 160619881-160619881 on Build GRCh38
rs766417929 Chr.1: 160619811-160619811 on Build GRCh38
rs761752439 Chr.1: 160619838-160619838 on Build GRCh38
rs556945293 Chr.1: 160619777-160619777 on Build GRCh38
rs904458133 Chr.1: 160619797-160619797 on Build GRCh38
rs754957258 Chr.1: 160619731-160619731 on Build GRCh38
rs183474893 Chr.1: 160619888-160619888 on Build GRCh38
rs756930532 Chr.1: 160619887-160619887 on Build GRCh38
rs998439405 Chr.1: 160619802-160619802 on Build GRCh38
rs752284590 Chr.1: 160619879-160619879 on Build GRCh38
rs201756489 Chr.1: 160619746-160619746 on Build GRCh38
rs780886162 Chr.1: 160619740-160619740 on Build GRCh38
rs1042950885 Chr.1: 160619772-160619772 on Build GRCh38
rs751442558 Chr.1: 160619810-160619811 on Build GRCh38
rs918515709 Chr.1: 160619915-160619915 on Build GRCh38
rs755812702 Chr.1: 160619744-160619744 on Build GRCh38
rs766339910 Chr.1: 160619847-160619847 on Build GRCh38
rs868160016 Chr.1: 160619787-160619787 on Build GRCh38
rs754473636 Chr.1: 160619713-160619713 on Build GRCh38
rs990341137 Chr.1: 160619747-160619747 on Build GRCh38
rs777248654 Chr.1: 160619761-160619761 on Build GRCh38
rs931107414 Chr.1: 160619926-160619926 on Build GRCh38
rs370951485 Chr.1: 160619766-160619766 on Build GRCh38
rs752231569 Chr.1: 160619743-160619743 on Build GRCh38
rs777495856 Chr.1: 160619886-160619886 on Build GRCh38
rs138438094 Chr.1: 160619885-160619885 on Build GRCh38
rs780384712 Chr.1: 160619780-160619780 on Build GRCh38
rs763221350 Chr.1: 160619846-160619846 on Build GRCh38
rs776744716 Chr.1: 160619826-160619826 on Build GRCh38
rs770088901 Chr.1: 160619839-160619839 on Build GRCh38
rs751364777 Chr.1: 160619855-160619855 on Build GRCh38
rs970133490 Chr.1: 160619831-160619831 on Build GRCh38

View Full Product Details