Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31580421-31580668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRD5A2
Entrez Gene ID
6716
Gene Name
steroid 5 alpha-reductase 2
Gene Aliases
-
UniGene
Hs.458345
Gene Chromosome Location
Chr.2: 31522486-31665651 on Build GRCh38
Transcripts
NM_000348.3 XM_011533072.2

SNP Detail

SNP ID Chromosome Location
rs763934279 Chr.2: 31580634-31580634 on Build GRCh38
rs61750389 Chr.2: 31580643-31580643 on Build GRCh38
rs867198056 Chr.2: 31580637-31580637 on Build GRCh38
rs904274646 Chr.2: 31580646-31580646 on Build GRCh38
rs895644192 Chr.2: 31580451-31580451 on Build GRCh38
rs999806016 Chr.2: 31580495-31580495 on Build GRCh38
rs1031334872 Chr.2: 31580497-31580497 on Build GRCh38
rs61750391 Chr.2: 31580607-31580607 on Build GRCh38
rs895624262 Chr.2: 31580625-31580625 on Build GRCh38
rs960511699 Chr.2: 31580449-31580450 on Build GRCh38
rs955059202 Chr.2: 31580554-31580554 on Build GRCh38
rs577592928 Chr.2: 31580542-31580542 on Build GRCh38
rs956950788 Chr.2: 31580499-31580499 on Build GRCh38
rs986338860 Chr.2: 31580556-31580556 on Build GRCh38
rs952022145 Chr.2: 31580488-31580488 on Build GRCh38
rs558048362 Chr.2: 31580506-31580506 on Build GRCh38
rs61750390 Chr.2: 31580628-31580628 on Build GRCh38
rs522638 Chr.2: 31580605-31580605 on Build GRCh38
rs752924894 Chr.2: 31580465-31580465 on Build GRCh38
rs752816270 Chr.2: 31580595-31580595 on Build GRCh38
rs758194712 Chr.2: 31580509-31580509 on Build GRCh38
rs947621822 Chr.2: 31580593-31580593 on Build GRCh38
rs555733249 Chr.2: 31580441-31580441 on Build GRCh38
rs191322008 Chr.2: 31580454-31580454 on Build GRCh38
rs747326745 Chr.2: 31580619-31580619 on Build GRCh38
rs619857 Chr.2: 31580549-31580549 on Build GRCh38
rs774959761 Chr.2: 31580609-31580609 on Build GRCh38
rs756975134 Chr.2: 31580645-31580645 on Build GRCh38
rs771649794 Chr.2: 31580555-31580555 on Build GRCh38
rs1012013692 Chr.2: 31580461-31580461 on Build GRCh38
rs754948149 Chr.2: 31580587-31580587 on Build GRCh38
rs942125187 Chr.2: 31580590-31580590 on Build GRCh38
rs523349 Chr.2: 31580636-31580636 on Build GRCh38
rs757173295 Chr.2: 31580599-31580599 on Build GRCh38
rs751702605 Chr.2: 31580585-31580585 on Build GRCh38
rs1023156943 Chr.2: 31580468-31580468 on Build GRCh38
rs61750393 Chr.2: 31580597-31580597 on Build GRCh38
rs904307280 Chr.2: 31580477-31580477 on Build GRCh38
rs61750394 Chr.2: 31580583-31580583 on Build GRCh38
rs1030465983 Chr.2: 31580539-31580539 on Build GRCh38
rs575441052 Chr.2: 31580445-31580445 on Build GRCh38
rs778850680 Chr.2: 31580648-31580648 on Build GRCh38
rs763356396 Chr.2: 31580603-31580603 on Build GRCh38
rs368886184 Chr.2: 31580602-31580602 on Build GRCh38
rs961824599 Chr.2: 31580507-31580507 on Build GRCh38
rs929203814 Chr.2: 31580606-31580606 on Build GRCh38
rs61750392 Chr.2: 31580600-31580600 on Build GRCh38
rs201175894 Chr.2: 31580630-31580630 on Build GRCh38
rs756284261 Chr.2: 31580631-31580631 on Build GRCh38
rs775228331 Chr.2: 31580601-31580601 on Build GRCh38
rs975810669 Chr.2: 31580537-31580537 on Build GRCh38
rs868416832 Chr.2: 31580638-31580638 on Build GRCh38
rs781510075 Chr.2: 31580594-31580594 on Build GRCh38
rs777512867 Chr.2: 31580598-31580598 on Build GRCh38

View Full Product Details