Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67727034-67727259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
XM_017020925.1 NM_152443.2

Gene Symbol
RDH12
Entrez Gene ID
145226
Gene Name
retinol dehydrogenase 12 (all-trans/9-cis/11-cis)
Gene Aliases
LCA13, RP53, SDR7C2
UniGene
Hs.415322
Gene Chromosome Location
Chr.14: 67701886-67734451 on Build GRCh38
Transcripts
XM_017020925.1 NM_152443.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890000 c.506G>C p.R169P Chr.14: 67727038-67727038 on Build GRCh38
6006113 c.565C>A p.Q189K Chr.14: 67727097-67727097 on Build GRCh38
4051966 c.578G>A p.R193H Chr.14: 67727110-67727110 on Build GRCh38
3497281 c.522C>T p.S174S Chr.14: 67727054-67727054 on Build GRCh38
4793690 c.618C>T p.A206A Chr.14: 67727150-67727150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998898101 Chr.14: 67727173-67727173 on Build GRCh38
rs104894472 Chr.14: 67727055-67727055 on Build GRCh38
rs116733939 Chr.14: 67727056-67727056 on Build GRCh38
rs146653515 Chr.14: 67727166-67727166 on Build GRCh38
rs753959716 Chr.14: 67727141-67727141 on Build GRCh38
rs368971478 Chr.14: 67727204-67727204 on Build GRCh38
rs574136093 Chr.14: 67727208-67727208 on Build GRCh38
rs144793387 Chr.14: 67727103-67727103 on Build GRCh38
rs116465785 Chr.14: 67727090-67727090 on Build GRCh38
rs780078705 Chr.14: 67727154-67727154 on Build GRCh38
rs115356583 Chr.14: 67727091-67727091 on Build GRCh38
rs756315539 Chr.14: 67727057-67727057 on Build GRCh38
rs567973980 Chr.14: 67727110-67727110 on Build GRCh38
rs878853341 Chr.14: 67727180-67727210 on Build GRCh38
rs1009028414 Chr.14: 67727137-67727137 on Build GRCh38
rs764346970 Chr.14: 67727222-67727222 on Build GRCh38
rs148629905 Chr.14: 67727109-67727109 on Build GRCh38
rs370741042 Chr.14: 67727054-67727054 on Build GRCh38
rs373032226 Chr.14: 67727062-67727062 on Build GRCh38
rs764600869 Chr.14: 67727108-67727108 on Build GRCh38
rs768588551 Chr.14: 67727120-67727120 on Build GRCh38
rs376350022 Chr.14: 67727087-67727087 on Build GRCh38
rs867508225 Chr.14: 67727147-67727147 on Build GRCh38
rs753074829 Chr.14: 67727114-67727114 on Build GRCh38
rs768529116 Chr.14: 67727167-67727167 on Build GRCh38
rs142117351 Chr.14: 67727111-67727111 on Build GRCh38
rs747732827 Chr.14: 67727182-67727182 on Build GRCh38
rs747183193 Chr.14: 67727153-67727153 on Build GRCh38
rs761052236 Chr.14: 67727213-67727213 on Build GRCh38
rs771268615 Chr.14: 67727162-67727162 on Build GRCh38
rs867284235 Chr.14: 67727122-67727122 on Build GRCh38
rs1032999368 Chr.14: 67727203-67727203 on Build GRCh38
rs762391730 Chr.14: 67727197-67727197 on Build GRCh38
rs775937640 Chr.14: 67727206-67727206 on Build GRCh38
rs372346477 Chr.14: 67727168-67727168 on Build GRCh38
rs371107041 Chr.14: 67727101-67727101 on Build GRCh38
rs140895705 Chr.14: 67727068-67727068 on Build GRCh38
rs535474444 Chr.14: 67727132-67727132 on Build GRCh38
rs776412935 Chr.14: 67727126-67727126 on Build GRCh38
rs769384787 Chr.14: 67727183-67727183 on Build GRCh38
rs891125540 Chr.14: 67727179-67727179 on Build GRCh38
rs104894470 Chr.14: 67727097-67727097 on Build GRCh38
rs772593066 Chr.14: 67727188-67727188 on Build GRCh38
rs368133299 Chr.14: 67727144-67727144 on Build GRCh38
rs778933833 Chr.14: 67727072-67727072 on Build GRCh38
rs387906272 Chr.14: 67727191-67727191 on Build GRCh38
rs745871149 Chr.14: 67727151-67727151 on Build GRCh38
rs374403088 Chr.14: 67727074-67727074 on Build GRCh38
rs756405179 Chr.14: 67727121-67727121 on Build GRCh38
rs79869437 Chr.14: 67727102-67727102 on Build GRCh38

View Full Product Details