Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 52084521-52084780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP64
Entrez Gene ID
55734
Gene Name
ZFP64 zinc finger protein
Gene Aliases
ZNF338
UniGene
Hs.473082
Gene Chromosome Location
Chr.20: 52084011-52191985 on Build GRCh38
Transcripts
NM_001319146.1 NM_199427.2 XM_006723822.1 XM_017027945.1 XM_017027947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841298 c.1797G>C p.K599N Chr.20: 52084698-52084698 on Build GRCh38
6826784 c.1767C>T p.F589F Chr.20: 52084728-52084728 on Build GRCh38
1233411 c.1870G>A p.G624R Chr.20: 52084625-52084625 on Build GRCh38
3770845 c.1783C>T p.L595L Chr.20: 52084712-52084712 on Build GRCh38
6826782 c.1722C>A p.I574I Chr.20: 52084773-52084773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747079054 Chr.20: 52084707-52084707 on Build GRCh38
rs750823485 Chr.20: 52084753-52084753 on Build GRCh38
rs779950515 Chr.20: 52084704-52084704 on Build GRCh38
rs753074279 Chr.20: 52084590-52084590 on Build GRCh38
rs752271185 Chr.20: 52084642-52084642 on Build GRCh38
rs771464973 Chr.20: 52084600-52084600 on Build GRCh38
rs751279449 Chr.20: 52084578-52084578 on Build GRCh38
rs763759145 Chr.20: 52084731-52084731 on Build GRCh38
rs746539783 Chr.20: 52084600-52084601 on Build GRCh38
rs759058553 Chr.20: 52084724-52084724 on Build GRCh38
rs778396467 Chr.20: 52084661-52084661 on Build GRCh38
rs766894959 Chr.20: 52084641-52084641 on Build GRCh38
rs777373723 Chr.20: 52084555-52084555 on Build GRCh38
rs769747430 Chr.20: 52084618-52084618 on Build GRCh38
rs772372079 Chr.20: 52084691-52084691 on Build GRCh38
rs754467359 Chr.20: 52084678-52084678 on Build GRCh38
rs6021695 Chr.20: 52084615-52084615 on Build GRCh38
rs754342890 Chr.20: 52084732-52084732 on Build GRCh38
rs764876176 Chr.20: 52084547-52084547 on Build GRCh38
rs201198421 Chr.20: 52084602-52084602 on Build GRCh38
rs569117649 Chr.20: 52084680-52084680 on Build GRCh38
rs752336058 Chr.20: 52084727-52084727 on Build GRCh38
rs754432281 Chr.20: 52084587-52084587 on Build GRCh38
rs746010569 Chr.20: 52084689-52084689 on Build GRCh38
rs761617382 Chr.20: 52084617-52084617 on Build GRCh38
rs765657938 Chr.20: 52084740-52084740 on Build GRCh38
rs756672726 Chr.20: 52084592-52084592 on Build GRCh38
rs144578934 Chr.20: 52084754-52084754 on Build GRCh38
rs768789242 Chr.20: 52084710-52084710 on Build GRCh38
rs759226203 Chr.20: 52084640-52084640 on Build GRCh38
rs375797185 Chr.20: 52084717-52084717 on Build GRCh38
rs773172109 Chr.20: 52084619-52084619 on Build GRCh38
rs990282830 Chr.20: 52084582-52084582 on Build GRCh38
rs773119259 Chr.20: 52084714-52084714 on Build GRCh38
rs755631260 Chr.20: 52084655-52084655 on Build GRCh38
rs750251656 Chr.20: 52084548-52084548 on Build GRCh38
rs140821732 Chr.20: 52084626-52084626 on Build GRCh38
rs552103606 Chr.20: 52084627-52084627 on Build GRCh38
rs770541250 Chr.20: 52084635-52084649 on Build GRCh38
rs779434068 Chr.20: 52084688-52084688 on Build GRCh38
rs775644020 Chr.20: 52084616-52084616 on Build GRCh38
rs751150706 Chr.20: 52084635-52084635 on Build GRCh38
rs373217247 Chr.20: 52084555-52084555 on Build GRCh38
rs765983017 Chr.20: 52084560-52084560 on Build GRCh38
rs770643497 Chr.20: 52084720-52084720 on Build GRCh38
rs772487454 Chr.20: 52084677-52084677 on Build GRCh38
rs142930891 Chr.20: 52084589-52084589 on Build GRCh38
rs775859440 Chr.20: 52084739-52084739 on Build GRCh38
rs767172743 Chr.20: 52084726-52084726 on Build GRCh38
rs757844138 Chr.20: 52084737-52084737 on Build GRCh38
rs150634112 Chr.20: 52084611-52084611 on Build GRCh38
rs749651360 Chr.20: 52084596-52084596 on Build GRCh38
rs939013265 Chr.20: 52084709-52084709 on Build GRCh38

View Full Product Details