Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18269650-18269919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAA1
Entrez Gene ID
6288
Gene Name
serum amyloid A1
Gene Aliases
PIG4, SAA, SAA2, TP53I4
UniGene
Hs.632144
Gene Chromosome Location
Chr.11: 18266225-18269977 on Build GRCh38
Transcripts
NM_000331.5 NM_001178006.2 NM_199161.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4845844 c.239G>C p.R80T Chr.11: 18269725-18269725 on Build GRCh38
284757 c.258C>A p.F86L Chr.11: 18269744-18269744 on Build GRCh38
4723379 c.232G>A p.D78N Chr.11: 18269718-18269718 on Build GRCh38
1637788 c.273G>A p.A91A Chr.11: 18269759-18269759 on Build GRCh38
3445979 c.295G>A p.A99T Chr.11: 18269781-18269781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767154283 Chr.11: 18269827-18269827 on Build GRCh38
rs11545465 Chr.11: 18269768-18269768 on Build GRCh38
rs772269963 Chr.11: 18269788-18269788 on Build GRCh38
rs547665021 Chr.11: 18269684-18269684 on Build GRCh38
rs374676694 Chr.11: 18269697-18269697 on Build GRCh38
rs368358385 Chr.11: 18269729-18269729 on Build GRCh38
rs371577716 Chr.11: 18269759-18269759 on Build GRCh38
rs1059559 Chr.11: 18269742-18269742 on Build GRCh38
rs771332080 Chr.11: 18269722-18269722 on Build GRCh38
rs751937285 Chr.11: 18269694-18269696 on Build GRCh38
rs879141095 Chr.11: 18269712-18269713 on Build GRCh38
rs774773641 Chr.11: 18269783-18269783 on Build GRCh38
rs746094081 Chr.11: 18269784-18269784 on Build GRCh38
rs61745680 Chr.11: 18269848-18269848 on Build GRCh38
rs183981346 Chr.11: 18269885-18269885 on Build GRCh38
rs1059571 Chr.11: 18269809-18269809 on Build GRCh38
rs756520340 Chr.11: 18269830-18269830 on Build GRCh38
rs17850140 Chr.11: 18269787-18269787 on Build GRCh38
rs557915415 Chr.11: 18269718-18269718 on Build GRCh38
rs761619125 Chr.11: 18269798-18269798 on Build GRCh38
rs753055758 Chr.11: 18269829-18269829 on Build GRCh38
rs2229338 Chr.11: 18269752-18269752 on Build GRCh38
rs15790 Chr.11: 18269765-18269765 on Build GRCh38
rs879202864 Chr.11: 18269712-18269712 on Build GRCh38
rs138257225 Chr.11: 18269801-18269801 on Build GRCh38
rs1059560 Chr.11: 18269746-18269746 on Build GRCh38
rs781421221 Chr.11: 18269766-18269766 on Build GRCh38
rs144465925 Chr.11: 18269745-18269745 on Build GRCh38
rs12218 Chr.11: 18269774-18269774 on Build GRCh38
rs776323374 Chr.11: 18269793-18269793 on Build GRCh38
rs765859609 Chr.11: 18269825-18269825 on Build GRCh38
rs547627072 Chr.11: 18269878-18269878 on Build GRCh38
rs150951768 Chr.11: 18269767-18269767 on Build GRCh38
rs142961925 Chr.11: 18269826-18269826 on Build GRCh38
rs778492890 Chr.11: 18269840-18269840 on Build GRCh38
rs751165905 Chr.11: 18269763-18269763 on Build GRCh38
rs573855702 Chr.11: 18269747-18269747 on Build GRCh38
rs770274549 Chr.11: 18269710-18269710 on Build GRCh38
rs764763274 Chr.11: 18269680-18269680 on Build GRCh38
rs559587106 Chr.11: 18269758-18269758 on Build GRCh38
rs774685920 Chr.11: 18269734-18269734 on Build GRCh38
rs879029191 Chr.11: 18269711-18269711 on Build GRCh38
rs145680768 Chr.11: 18269773-18269773 on Build GRCh38
rs113832815 Chr.11: 18269713-18269713 on Build GRCh38
rs779633638 Chr.11: 18269674-18269674 on Build GRCh38
rs747421710 Chr.11: 18269675-18269675 on Build GRCh38
rs545246718 Chr.11: 18269781-18269781 on Build GRCh38
rs749956999 Chr.11: 18269756-18269756 on Build GRCh38
rs1059567 Chr.11: 18269790-18269790 on Build GRCh38
rs79681911 Chr.11: 18269755-18269755 on Build GRCh38
rs539813336 Chr.11: 18269717-18269717 on Build GRCh38
rs148453200 Chr.11: 18269808-18269809 on Build GRCh38
rs779140717 Chr.11: 18269852-18269852 on Build GRCh38
rs985505512 Chr.11: 18269814-18269814 on Build GRCh38
rs376950598 Chr.11: 18269670-18269670 on Build GRCh38
rs762516122 Chr.11: 18269811-18269811 on Build GRCh38
rs748587828 Chr.11: 18269708-18269708 on Build GRCh38

View Full Product Details