Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152155808-152156071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPTN
Entrez Gene ID
126638
Gene Name
repetin
Gene Aliases
-
UniGene
Hs.376144
Gene Chromosome Location
Chr.1: 152153595-152159228 on Build GRCh38
Transcripts
NM_001122965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6280093 c.1035A>G p.Q345Q Chr.1: 152156064-152156064 on Build GRCh38
5037810 c.1221G>C p.E407D Chr.1: 152155878-152155878 on Build GRCh38
3930400 c.1211G>A p.G404D Chr.1: 152155888-152155888 on Build GRCh38
2184370 c.1238C>T p.S413F Chr.1: 152155861-152155861 on Build GRCh38
5843659 c.1096C>T p.H366Y Chr.1: 152156003-152156003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778667714 Chr.1: 152155927-152155927 on Build GRCh38
rs1034712371 Chr.1: 152155938-152155938 on Build GRCh38
rs374167664 Chr.1: 152155878-152155878 on Build GRCh38
rs767585803 Chr.1: 152155977-152155977 on Build GRCh38
rs200503098 Chr.1: 152155925-152155925 on Build GRCh38
rs563014612 Chr.1: 152155992-152155992 on Build GRCh38
rs779574736 Chr.1: 152156015-152156015 on Build GRCh38
rs202018143 Chr.1: 152156037-152156037 on Build GRCh38
rs766633550 Chr.1: 152155968-152155968 on Build GRCh38
rs750415348 Chr.1: 152155987-152155987 on Build GRCh38
rs753697715 Chr.1: 152155978-152155978 on Build GRCh38
rs773952962 Chr.1: 152155834-152155834 on Build GRCh38
rs574189458 Chr.1: 152155981-152155981 on Build GRCh38
rs777079905 Chr.1: 152155940-152155940 on Build GRCh38
rs756933648 Chr.1: 152155982-152155982 on Build GRCh38
rs181516672 Chr.1: 152155942-152155942 on Build GRCh38
rs377116556 Chr.1: 152155994-152155994 on Build GRCh38
rs762417902 Chr.1: 152155853-152155853 on Build GRCh38
rs768176912 Chr.1: 152155936-152155936 on Build GRCh38
rs756306102 Chr.1: 152155879-152155914 on Build GRCh38
rs200109397 Chr.1: 152155946-152155946 on Build GRCh38
rs905642327 Chr.1: 152155874-152155874 on Build GRCh38
rs1011497967 Chr.1: 152155900-152155900 on Build GRCh38
rs201910869 Chr.1: 152155894-152155894 on Build GRCh38
rs201068375 Chr.1: 152155937-152155937 on Build GRCh38
rs377399394 Chr.1: 152155950-152155950 on Build GRCh38
rs370044683 Chr.1: 152155896-152155896 on Build GRCh38
rs778921335 Chr.1: 152155984-152155984 on Build GRCh38
rs765618135 Chr.1: 152155855-152155855 on Build GRCh38
rs768389362 Chr.1: 152156039-152156039 on Build GRCh38
rs1047336143 Chr.1: 152155973-152155973 on Build GRCh38
rs370015842 Chr.1: 152155933-152155933 on Build GRCh38
rs776850079 Chr.1: 152155852-152155852 on Build GRCh38
rs200646250 Chr.1: 152155832-152155832 on Build GRCh38
rs758324037 Chr.1: 152156002-152156002 on Build GRCh38
rs567965937 Chr.1: 152155888-152155888 on Build GRCh38
rs751896102 Chr.1: 152155873-152155873 on Build GRCh38
rs749006751 Chr.1: 152155905-152155905 on Build GRCh38
rs750535812 Chr.1: 152155857-152155857 on Build GRCh38
rs771510334 Chr.1: 152155930-152155930 on Build GRCh38
rs761258474 Chr.1: 152155919-152155919 on Build GRCh38
rs746477927 Chr.1: 152156034-152156034 on Build GRCh38
rs758492530 Chr.1: 152155863-152155863 on Build GRCh38
rs200041751 Chr.1: 152155988-152155988 on Build GRCh38
rs763542756 Chr.1: 152155951-152155951 on Build GRCh38
rs538216535 Chr.1: 152155841-152155841 on Build GRCh38
rs745342849 Chr.1: 152155835-152155835 on Build GRCh38
rs532048394 Chr.1: 152156009-152156009 on Build GRCh38
rs761035842 Chr.1: 152155846-152155846 on Build GRCh38
rs964100200 Chr.1: 152156045-152156045 on Build GRCh38
rs199724746 Chr.1: 152155870-152155870 on Build GRCh38
rs1038445603 Chr.1: 152155934-152155934 on Build GRCh38
rs751659467 Chr.1: 152155972-152155972 on Build GRCh38
rs764513210 Chr.1: 152155848-152155848 on Build GRCh38

View Full Product Details