Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114672131-114672372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX3
Entrez Gene ID
6926
Gene Name
T-box 3
Gene Aliases
TBX3-ISO, UMS, XHL
UniGene
Hs.744016
Gene Chromosome Location
Chr.12: 114670254-114684164 on Build GRCh38
Transcripts
NM_005996.3 NM_016569.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6917208 c.1787C>T p.P596L Chr.12: 114672286-114672286 on Build GRCh38
1127548 c.1843T>G p.S615A Chr.12: 114672230-114672230 on Build GRCh38
4918852 c.1897A>T p.T633S Chr.12: 114672176-114672176 on Build GRCh38
6917207 c.1810C>T p.P604S Chr.12: 114672263-114672263 on Build GRCh38
6944192 c.1876delC p.H626fs*6 Chr.12: 114672197-114672197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771698419 Chr.12: 114672203-114672203 on Build GRCh38
rs759202204 Chr.12: 114672202-114672202 on Build GRCh38
rs200821102 Chr.12: 114672223-114672223 on Build GRCh38
rs777552683 Chr.12: 114672186-114672186 on Build GRCh38
rs1040183921 Chr.12: 114672177-114672177 on Build GRCh38
rs774983428 Chr.12: 114672325-114672325 on Build GRCh38
rs751628989 Chr.12: 114672245-114672245 on Build GRCh38
rs762052798 Chr.12: 114672216-114672216 on Build GRCh38
rs189469110 Chr.12: 114672175-114672175 on Build GRCh38
rs777583851 Chr.12: 114672268-114672268 on Build GRCh38
rs763565421 Chr.12: 114672337-114672337 on Build GRCh38
rs755884541 Chr.12: 114672256-114672256 on Build GRCh38
rs753940578 Chr.12: 114672282-114672282 on Build GRCh38
rs773785943 Chr.12: 114672200-114672200 on Build GRCh38
rs766569858 Chr.12: 114672242-114672242 on Build GRCh38
rs971835561 Chr.12: 114672263-114672263 on Build GRCh38
rs1044589578 Chr.12: 114672342-114672342 on Build GRCh38
rs370501220 Chr.12: 114672319-114672319 on Build GRCh38
rs528719787 Chr.12: 114672213-114672213 on Build GRCh38
rs749188927 Chr.12: 114672278-114672278 on Build GRCh38
rs765821877 Chr.12: 114672221-114672221 on Build GRCh38
rs993513286 Chr.12: 114672234-114672234 on Build GRCh38
rs867417931 Chr.12: 114672289-114672289 on Build GRCh38
rs752590045 Chr.12: 114672255-114672255 on Build GRCh38
rs781193551 Chr.12: 114672170-114672170 on Build GRCh38
rs985763089 Chr.12: 114672308-114672308 on Build GRCh38
rs747894358 Chr.12: 114672181-114672181 on Build GRCh38
rs778450474 Chr.12: 114672283-114672283 on Build GRCh38
rs958925981 Chr.12: 114672231-114672231 on Build GRCh38
rs755321111 Chr.12: 114672251-114672251 on Build GRCh38
rs867049310 Chr.12: 114672241-114672241 on Build GRCh38
rs763473739 Chr.12: 114672230-114672230 on Build GRCh38
rs199825159 Chr.12: 114672328-114672328 on Build GRCh38
rs181219227 Chr.12: 114672346-114672346 on Build GRCh38
rs954634731 Chr.12: 114672195-114672195 on Build GRCh38
rs770459748 Chr.12: 114672198-114672198 on Build GRCh38
rs769814877 Chr.12: 114672182-114672182 on Build GRCh38
rs3186021 Chr.12: 114672187-114672187 on Build GRCh38
rs374540986 Chr.12: 114672338-114672338 on Build GRCh38
rs755181770 Chr.12: 114672166-114672166 on Build GRCh38
rs930385721 Chr.12: 114672290-114672290 on Build GRCh38
rs1030153498 Chr.12: 114672154-114672154 on Build GRCh38
rs946996946 Chr.12: 114672227-114672227 on Build GRCh38

View Full Product Details