Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100085179-100085416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SASS6
Entrez Gene ID
163786
Gene Name
SAS-6 centriolar assembly protein
Gene Aliases
MCPH14, SAS-6, SAS6
UniGene
Hs.591447
Gene Chromosome Location
Chr.1: 100082632-100133095 on Build GRCh38
Transcripts
NM_001304829.1 NM_194292.2 XM_017000486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6445251 c.1961T>A p.L654* Chr.1: 100085341-100085341 on Build GRCh38
4600393 c.1918A>G p.T640A Chr.1: 100085384-100085384 on Build GRCh38
5842860 c.1911delC p.K638fs*>20 Chr.1: 100085391-100085391 on Build GRCh38
4974512 c.1935G>A p.A645A Chr.1: 100085367-100085367 on Build GRCh38
3470283 c.1921G>A p.A641T Chr.1: 100085381-100085381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756409319 Chr.1: 100085311-100085311 on Build GRCh38
rs936662083 Chr.1: 100085289-100085289 on Build GRCh38
rs760754724 Chr.1: 100085373-100085373 on Build GRCh38
rs781637620 Chr.1: 100085303-100085303 on Build GRCh38
rs12039960 Chr.1: 100085283-100085283 on Build GRCh38
rs770629923 Chr.1: 100085379-100085379 on Build GRCh38
rs763067017 Chr.1: 100085386-100085386 on Build GRCh38
rs190291928 Chr.1: 100085275-100085275 on Build GRCh38
rs901693179 Chr.1: 100085387-100085387 on Build GRCh38
rs751713581 Chr.1: 100085280-100085280 on Build GRCh38
rs1054012011 Chr.1: 100085346-100085346 on Build GRCh38
rs773227677 Chr.1: 100085371-100085371 on Build GRCh38
rs139681683 Chr.1: 100085368-100085368 on Build GRCh38
rs778766058 Chr.1: 100085350-100085350 on Build GRCh38
rs933391974 Chr.1: 100085222-100085222 on Build GRCh38
rs748886279 Chr.1: 100085338-100085338 on Build GRCh38
rs927085671 Chr.1: 100085335-100085335 on Build GRCh38
rs1040283628 Chr.1: 100085385-100085385 on Build GRCh38
rs144990398 Chr.1: 100085367-100085367 on Build GRCh38
rs768393413 Chr.1: 100085357-100085357 on Build GRCh38
rs780147019 Chr.1: 100085210-100085211 on Build GRCh38
rs878916312 Chr.1: 100085261-100085261 on Build GRCh38
rs750929575 Chr.1: 100085304-100085304 on Build GRCh38
rs778264546 Chr.1: 100085361-100085361 on Build GRCh38
rs757352869 Chr.1: 100085282-100085282 on Build GRCh38
rs148677150 Chr.1: 100085381-100085381 on Build GRCh38
rs923897617 Chr.1: 100085268-100085268 on Build GRCh38
rs780523206 Chr.1: 100085330-100085330 on Build GRCh38
rs144334982 Chr.1: 100085243-100085243 on Build GRCh38

View Full Product Details