Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139539515-139539786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507291
Entrez Gene ID
100507291
Gene Name
uncharacterized LOC100507291
Gene Aliases
-
UniGene
Hs.699071
Gene Chromosome Location
Chr.3: 139389803-139583319 on Build GRCh38
Transcripts
NR_121609.1 NM_001130992.1 NM_001130993.1 NM_002899.3

Gene Symbol
RBP1
Entrez Gene ID
5947
Gene Name
retinol binding protein 1
Gene Aliases
CRABP-I, CRBP, CRBP1, CRBPI, RBPC
UniGene
Hs.529571
Gene Chromosome Location
Chr.3: 139517434-139539829 on Build GRCh38
Transcripts
NR_121609.1 NM_001130992.1 NM_001130993.1 NM_002899.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5166874 c.33_34insGG p.N12fs*89 Chr.3: 139539685-139539686 on Build GRCh38
1484675 c.196G>A p.D66N Chr.3: 139539523-139539523 on Build GRCh38
4813080 c.10C>T p.P4S Chr.3: 139539709-139539709 on Build GRCh38
4628145 c.200T>C p.F67S Chr.3: 139539519-139539519 on Build GRCh38
4628146 c.200T>C p.F67S Chr.3: 139539519-139539519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746224991 Chr.3: 139539640-139539640 on Build GRCh38
rs372765878 Chr.3: 139539755-139539755 on Build GRCh38
rs572591434 Chr.3: 139539593-139539593 on Build GRCh38
rs771081681 Chr.3: 139539617-139539617 on Build GRCh38
rs765950525 Chr.3: 139539679-139539679 on Build GRCh38
rs11549984 Chr.3: 139539569-139539569 on Build GRCh38
rs1038375447 Chr.3: 139539540-139539540 on Build GRCh38
rs759895995 Chr.3: 139539559-139539559 on Build GRCh38
rs749326153 Chr.3: 139539615-139539615 on Build GRCh38
rs968254145 Chr.3: 139539647-139539647 on Build GRCh38
rs781124300 Chr.3: 139539543-139539543 on Build GRCh38
rs749489464 Chr.3: 139539551-139539551 on Build GRCh38
rs777799194 Chr.3: 139539609-139539609 on Build GRCh38
rs761484938 Chr.3: 139539573-139539573 on Build GRCh38
rs561177753 Chr.3: 139539674-139539674 on Build GRCh38
rs748227827 Chr.3: 139539545-139539545 on Build GRCh38
rs965761248 Chr.3: 139539739-139539739 on Build GRCh38
rs764828782 Chr.3: 139539576-139539576 on Build GRCh38
rs769611462 Chr.3: 139539540-139539540 on Build GRCh38
rs546485479 Chr.3: 139539653-139539653 on Build GRCh38
rs111548824 Chr.3: 139539748-139539748 on Build GRCh38
rs895860218 Chr.3: 139539554-139539554 on Build GRCh38
rs767797680 Chr.3: 139539655-139539655 on Build GRCh38
rs905311848 Chr.3: 139539567-139539567 on Build GRCh38
rs1036477406 Chr.3: 139539654-139539655 on Build GRCh38
rs953157042 Chr.3: 139539704-139539704 on Build GRCh38
rs183138806 Chr.3: 139539719-139539719 on Build GRCh38
rs545322916 Chr.3: 139539552-139539552 on Build GRCh38
rs758095834 Chr.3: 139539581-139539581 on Build GRCh38
rs2071386 Chr.3: 139539666-139539666 on Build GRCh38
rs1010119517 Chr.3: 139539598-139539598 on Build GRCh38
rs754785993 Chr.3: 139539692-139539692 on Build GRCh38
rs1044301751 Chr.3: 139539765-139539765 on Build GRCh38
rs559625725 Chr.3: 139539602-139539602 on Build GRCh38
rs750154057 Chr.3: 139539577-139539577 on Build GRCh38
rs766004286 Chr.3: 139539584-139539584 on Build GRCh38
rs774576614 Chr.3: 139539625-139539625 on Build GRCh38
rs922991645 Chr.3: 139539718-139539718 on Build GRCh38
rs975860398 Chr.3: 139539703-139539703 on Build GRCh38
rs368158259 Chr.3: 139539548-139539548 on Build GRCh38
rs774474421 Chr.3: 139539571-139539571 on Build GRCh38
rs761245937 Chr.3: 139539660-139539660 on Build GRCh38
rs756076216 Chr.3: 139539738-139539738 on Build GRCh38
rs769858847 Chr.3: 139539546-139539546 on Build GRCh38
rs754733560 Chr.3: 139539591-139539591 on Build GRCh38
rs751321164 Chr.3: 139539585-139539585 on Build GRCh38
rs767319794 Chr.3: 139539712-139539712 on Build GRCh38
rs530222014 Chr.3: 139539687-139539687 on Build GRCh38
rs754890093 Chr.3: 139539541-139539541 on Build GRCh38
rs781054524 Chr.3: 139539592-139539592 on Build GRCh38
rs941918729 Chr.3: 139539746-139539746 on Build GRCh38
rs1021189561 Chr.3: 139539605-139539605 on Build GRCh38
rs979108517 Chr.3: 139539654-139539654 on Build GRCh38
rs774611475 Chr.3: 139539555-139539555 on Build GRCh38
rs772867674 Chr.3: 139539673-139539673 on Build GRCh38
rs1007515633 Chr.3: 139539705-139539705 on Build GRCh38
rs776199461 Chr.3: 139539659-139539659 on Build GRCh38
rs1027491485 Chr.3: 139539702-139539702 on Build GRCh38
rs767311866 Chr.3: 139539686-139539686 on Build GRCh38
rs983670523 Chr.3: 139539735-139539735 on Build GRCh38
rs909166990 Chr.3: 139539742-139539742 on Build GRCh38
rs756233491 Chr.3: 139539594-139539594 on Build GRCh38

View Full Product Details