Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57419991-57420220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF17
Entrez Gene ID
7565
Gene Name
zinc finger protein 17
Gene Aliases
HPF3, KOX10
UniGene
Hs.729403
Gene Chromosome Location
Chr.19: 57409252-57421939 on Build GRCh38
Transcripts
NM_006959.2 XM_017027205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3404709 c.523A>G p.R175G Chr.19: 57420015-57420015 on Build GRCh38
6086315 c.726A>T p.G242G Chr.19: 57420218-57420218 on Build GRCh38
1397019 c.649C>T p.P217S Chr.19: 57420141-57420141 on Build GRCh38
3773195 c.528C>A p.D176E Chr.19: 57420020-57420020 on Build GRCh38
1002217 c.543G>T p.E181D Chr.19: 57420035-57420035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012605873 Chr.19: 57420114-57420114 on Build GRCh38
rs763552531 Chr.19: 57420105-57420105 on Build GRCh38
rs749384061 Chr.19: 57420024-57420024 on Build GRCh38
rs374905485 Chr.19: 57420188-57420188 on Build GRCh38
rs751232385 Chr.19: 57420106-57420106 on Build GRCh38
rs200572163 Chr.19: 57420017-57420017 on Build GRCh38
rs750819885 Chr.19: 57420138-57420138 on Build GRCh38
rs776060768 Chr.19: 57420018-57420018 on Build GRCh38
rs10417533 Chr.19: 57420185-57420185 on Build GRCh38
rs762080016 Chr.19: 57420052-57420052 on Build GRCh38
rs779185578 Chr.19: 57420176-57420176 on Build GRCh38
rs749397263 Chr.19: 57420048-57420057 on Build GRCh38
rs757911234 Chr.19: 57420102-57420102 on Build GRCh38
rs28418770 Chr.19: 57420057-57420057 on Build GRCh38
rs773474502 Chr.19: 57420187-57420187 on Build GRCh38
rs749663430 Chr.19: 57420068-57420068 on Build GRCh38
rs756948339 Chr.19: 57420116-57420116 on Build GRCh38
rs756487976 Chr.19: 57420151-57420151 on Build GRCh38
rs768880842 Chr.19: 57420167-57420167 on Build GRCh38
rs781484279 Chr.19: 57420014-57420014 on Build GRCh38
rs368071786 Chr.19: 57420128-57420128 on Build GRCh38
rs757647323 Chr.19: 57420012-57420012 on Build GRCh38
rs759375090 Chr.19: 57420070-57420070 on Build GRCh38
rs538372230 Chr.19: 57420166-57420166 on Build GRCh38
rs772489710 Chr.19: 57420186-57420186 on Build GRCh38
rs746600701 Chr.19: 57420010-57420010 on Build GRCh38
rs776331717 Chr.19: 57420058-57420058 on Build GRCh38
rs775014735 Chr.19: 57420130-57420130 on Build GRCh38
rs369885721 Chr.19: 57420033-57420033 on Build GRCh38
rs768797019 Chr.19: 57420028-57420028 on Build GRCh38
rs199697551 Chr.19: 57420043-57420043 on Build GRCh38
rs375032874 Chr.19: 57420074-57420074 on Build GRCh38
rs752625025 Chr.19: 57420092-57420092 on Build GRCh38
rs780437090 Chr.19: 57420164-57420164 on Build GRCh38

View Full Product Details