Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43523801-43524074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1C
Entrez Gene ID
9533
Gene Name
RNA polymerase I subunit C
Gene Aliases
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3
UniGene
Hs.743509
Gene Chromosome Location
Chr.6: 43517024-43562404 on Build GRCh38
Transcripts
NM_001318876.1 XM_011515000.2 NM_020750.2

Gene Symbol
XPO5
Entrez Gene ID
57510
Gene Name
exportin 5
Gene Aliases
exp5
UniGene
Hs.203206
Gene Chromosome Location
Chr.6: 43522330-43576075 on Build GRCh38
Transcripts
NM_001318876.1 XM_011515000.2 NM_020750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6823999 c.3595G>A p.A1199T Chr.6: 43523888-43523888 on Build GRCh38
1444741 c.3581A>G p.D1194G Chr.6: 43523902-43523902 on Build GRCh38
4589829 c.3545_3546insA p.P1184fs*11 Chr.6: 43523937-43523938 on Build GRCh38
5436719 c.3531A>G p.S1177S Chr.6: 43523952-43523952 on Build GRCh38
4611658 c.3544_3545insA p.T1182fs*13 Chr.6: 43523938-43523939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537877471 Chr.6: 43523884-43523884 on Build GRCh38
rs376380751 Chr.6: 43523913-43523913 on Build GRCh38
rs761029519 Chr.6: 43523835-43523835 on Build GRCh38
rs372044826 Chr.6: 43523835-43523835 on Build GRCh38
rs746915857 Chr.6: 43523853-43523853 on Build GRCh38
rs202110100 Chr.6: 43524020-43524020 on Build GRCh38
rs754211966 Chr.6: 43523965-43523965 on Build GRCh38
rs745552409 Chr.6: 43523876-43523876 on Build GRCh38
rs757103216 Chr.6: 43524041-43524041 on Build GRCh38
rs746980628 Chr.6: 43524039-43524039 on Build GRCh38
rs983939759 Chr.6: 43523983-43523983 on Build GRCh38
rs762491983 Chr.6: 43523919-43523919 on Build GRCh38
rs773520890 Chr.6: 43523833-43523835 on Build GRCh38
rs1037364083 Chr.6: 43523887-43523887 on Build GRCh38
rs755180111 Chr.6: 43523829-43523829 on Build GRCh38
rs777701567 Chr.6: 43524027-43524027 on Build GRCh38
rs755270020 Chr.6: 43523970-43523970 on Build GRCh38
rs970321067 Chr.6: 43523976-43523976 on Build GRCh38
rs373525815 Chr.6: 43523921-43523921 on Build GRCh38
rs761034663 Chr.6: 43523930-43523930 on Build GRCh38
rs766448717 Chr.6: 43524049-43524078 on Build GRCh38
rs758384615 Chr.6: 43523842-43523842 on Build GRCh38
rs758570217 Chr.6: 43524025-43524025 on Build GRCh38
rs765603626 Chr.6: 43523978-43523978 on Build GRCh38
rs369309281 Chr.6: 43523920-43523920 on Build GRCh38
rs776341368 Chr.6: 43523873-43523873 on Build GRCh38
rs778848443 Chr.6: 43523843-43523843 on Build GRCh38
rs770816576 Chr.6: 43523861-43523861 on Build GRCh38
rs766767634 Chr.6: 43523955-43523955 on Build GRCh38

View Full Product Details