Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89438470-89438743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A12
Entrez Gene ID
387700
Gene Name
solute carrier family 16 member 12
Gene Aliases
CJMG, CRT2, MCT12
UniGene
Hs.530338
Gene Chromosome Location
Chr.10: 89430294-89536028 on Build GRCh38
Transcripts
NM_213606.3 XM_017016237.1 XM_017016238.1 XM_017016239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551130 c.968C>A p.S323Y Chr.10: 89438664-89438664 on Build GRCh38
5515360 c.938+9C>A p.? Chr.10: 89438595-89438595 on Build GRCh38
3441403 c.858G>A p.Q286Q Chr.10: 89438684-89438684 on Build GRCh38
7141526 c.1002C>A p.I334I Chr.10: 89438630-89438630 on Build GRCh38
4932560 c.817T>C p.Y273H Chr.10: 89438725-89438725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758860646 Chr.10: 89438640-89438640 on Build GRCh38
rs370150531 Chr.10: 89438719-89438719 on Build GRCh38
rs999321057 Chr.10: 89438529-89438529 on Build GRCh38
rs750912507 Chr.10: 89438615-89438615 on Build GRCh38
rs904023445 Chr.10: 89438507-89438507 on Build GRCh38
rs770791449 Chr.10: 89438717-89438717 on Build GRCh38
rs1000225569 Chr.10: 89438550-89438550 on Build GRCh38
rs369583268 Chr.10: 89438559-89438559 on Build GRCh38
rs907771363 Chr.10: 89438680-89438680 on Build GRCh38
rs779542261 Chr.10: 89438612-89438612 on Build GRCh38
rs530890361 Chr.10: 89438496-89438496 on Build GRCh38
rs969757649 Chr.10: 89438586-89438586 on Build GRCh38
rs372825345 Chr.10: 89438565-89438565 on Build GRCh38
rs868353683 Chr.10: 89438614-89438614 on Build GRCh38
rs773018862 Chr.10: 89438676-89438676 on Build GRCh38
rs1019154259 Chr.10: 89438547-89438547 on Build GRCh38
rs148013353 Chr.10: 89438662-89438662 on Build GRCh38
rs940060935 Chr.10: 89438608-89438608 on Build GRCh38
rs556762831 Chr.10: 89438665-89438665 on Build GRCh38
rs762806269 Chr.10: 89438716-89438716 on Build GRCh38
rs780124609 Chr.10: 89438647-89438647 on Build GRCh38
rs756468882 Chr.10: 89438583-89438583 on Build GRCh38
rs751031196 Chr.10: 89438561-89438561 on Build GRCh38
rs1031235157 Chr.10: 89438555-89438555 on Build GRCh38
rs754253065 Chr.10: 89438583-89438583 on Build GRCh38
rs563387145 Chr.10: 89438594-89438594 on Build GRCh38
rs868663250 Chr.10: 89438646-89438646 on Build GRCh38
rs574208009 Chr.10: 89438575-89438575 on Build GRCh38
rs190510733 Chr.10: 89438652-89438652 on Build GRCh38
rs375233665 Chr.10: 89438570-89438570 on Build GRCh38

View Full Product Details