Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54320460-54320701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TINAG
Entrez Gene ID
27283
Gene Name
tubulointerstitial nephritis antigen
Gene Aliases
TIN-AG
UniGene
Hs.127011
Gene Chromosome Location
Chr.6: 54307792-54390152 on Build GRCh38
Transcripts
NM_014464.3 XM_006715062.2 XM_011514497.2 XM_017010745.1 XM_017010746.1 XM_017010747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3628623 c.313G>A p.D105N Chr.6: 54320590-54320590 on Build GRCh38
1697422 c.382G>A p.E128K Chr.6: 54320605-54320605 on Build GRCh38
3628622 c.355G>A p.D119N Chr.6: 54320590-54320590 on Build GRCh38
1697423 c.370G>A p.E124K Chr.6: 54320605-54320605 on Build GRCh38
6107401 c.419+1G>T p.? Chr.6: 54320643-54320643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750630178 Chr.6: 54320578-54320578 on Build GRCh38
rs748704001 Chr.6: 54320611-54320611 on Build GRCh38
rs999913872 Chr.6: 54320657-54320657 on Build GRCh38
rs774871147 Chr.6: 54320540-54320540 on Build GRCh38
rs770096004 Chr.6: 54320619-54320619 on Build GRCh38
rs945059969 Chr.6: 54320643-54320643 on Build GRCh38
rs571242627 Chr.6: 54320602-54320602 on Build GRCh38
rs573229339 Chr.6: 54320487-54320487 on Build GRCh38
rs1041384899 Chr.6: 54320612-54320612 on Build GRCh38
rs757134950 Chr.6: 54320567-54320567 on Build GRCh38
rs760070552 Chr.6: 54320660-54320660 on Build GRCh38
rs773597964 Chr.6: 54320635-54320635 on Build GRCh38
rs970997984 Chr.6: 54320508-54320508 on Build GRCh38
rs544499855 Chr.6: 54320555-54320555 on Build GRCh38
rs771639773 Chr.6: 54320641-54320641 on Build GRCh38
rs773971801 Chr.6: 54320537-54320537 on Build GRCh38
rs765143180 Chr.6: 54320571-54320571 on Build GRCh38
rs747044793 Chr.6: 54320599-54320599 on Build GRCh38
rs77785572 Chr.6: 54320519-54320519 on Build GRCh38
rs758698119 Chr.6: 54320579-54320579 on Build GRCh38
rs766979124 Chr.6: 54320539-54320539 on Build GRCh38
rs748079936 Chr.6: 54320619-54320619 on Build GRCh38
rs367701839 Chr.6: 54320640-54320640 on Build GRCh38
rs989644845 Chr.6: 54320529-54320529 on Build GRCh38
rs759208803 Chr.6: 54320538-54320538 on Build GRCh38
rs775273837 Chr.6: 54320644-54320644 on Build GRCh38
rs780122849 Chr.6: 54320584-54320584 on Build GRCh38
rs545499769 Chr.6: 54320517-54320517 on Build GRCh38
rs201925900 Chr.6: 54320548-54320548 on Build GRCh38
rs760605389 Chr.6: 54320546-54320546 on Build GRCh38
rs981013836 Chr.6: 54320513-54320513 on Build GRCh38
rs778915584 Chr.6: 54320610-54320610 on Build GRCh38
rs755149939 Chr.6: 54320608-54320608 on Build GRCh38
rs377487387 Chr.6: 54320662-54320662 on Build GRCh38

View Full Product Details