Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28957888-28958136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIL
Entrez Gene ID
9865
Gene Name
TLR4 interactor with leucine rich repeats
Gene Aliases
-
UniGene
Hs.744120
Gene Chromosome Location
Chr.7: 28953358-28958413 on Build GRCh38
Transcripts
NM_014817.3

SNP Detail

SNP ID Chromosome Location
rs759468605 Chr.7: 28957972-28957972 on Build GRCh38
rs750048675 Chr.7: 28958013-28958013 on Build GRCh38
rs775106481 Chr.7: 28958071-28958071 on Build GRCh38
rs767226561 Chr.7: 28958010-28958010 on Build GRCh38
rs867676921 Chr.7: 28958106-28958106 on Build GRCh38
rs775543492 Chr.7: 28957979-28957979 on Build GRCh38
rs740251 Chr.7: 28958075-28958075 on Build GRCh38
rs869306388 Chr.7: 28957982-28957983 on Build GRCh38
rs755385398 Chr.7: 28958014-28958014 on Build GRCh38
rs550555799 Chr.7: 28957911-28957911 on Build GRCh38
rs747118407 Chr.7: 28958035-28958035 on Build GRCh38
rs748581791 Chr.7: 28957947-28957947 on Build GRCh38
rs753283001 Chr.7: 28958023-28958023 on Build GRCh38
rs758768615 Chr.7: 28958024-28958024 on Build GRCh38
rs755333993 Chr.7: 28957908-28957908 on Build GRCh38
rs1056494644 Chr.7: 28958036-28958036 on Build GRCh38
rs549106202 Chr.7: 28958050-28958050 on Build GRCh38
rs747262957 Chr.7: 28957964-28957964 on Build GRCh38
rs752533945 Chr.7: 28957978-28957979 on Build GRCh38
rs772220595 Chr.7: 28957954-28957954 on Build GRCh38
rs762749511 Chr.7: 28957985-28957985 on Build GRCh38
rs750965799 Chr.7: 28958003-28958003 on Build GRCh38
rs769728795 Chr.7: 28958064-28958064 on Build GRCh38
rs776977176 Chr.7: 28957971-28957971 on Build GRCh38
rs754261518 Chr.7: 28957906-28957906 on Build GRCh38
rs777679064 Chr.7: 28957962-28957962 on Build GRCh38
rs1023598230 Chr.7: 28958074-28958074 on Build GRCh38
rs1025395390 Chr.7: 28957965-28957965 on Build GRCh38
rs777925831 Chr.7: 28958028-28958028 on Build GRCh38
rs201596275 Chr.7: 28958016-28958016 on Build GRCh38
rs567694141 Chr.7: 28958057-28958057 on Build GRCh38
rs1011086225 Chr.7: 28958081-28958081 on Build GRCh38
rs763690469 Chr.7: 28957990-28957990 on Build GRCh38
rs761466591 Chr.7: 28958004-28958004 on Build GRCh38
rs1043979827 Chr.7: 28957975-28957975 on Build GRCh38
rs764069685 Chr.7: 28958075-28958076 on Build GRCh38
rs771223479 Chr.7: 28957967-28957967 on Build GRCh38
rs779437553 Chr.7: 28957922-28957922 on Build GRCh38
rs896590766 Chr.7: 28958047-28958047 on Build GRCh38
rs869082108 Chr.7: 28957981-28957982 on Build GRCh38

View Full Product Details