Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43873427-43873660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF404
Entrez Gene ID
342908
Gene Name
zinc finger protein 404
Gene Aliases
-
UniGene
Hs.76561
Gene Chromosome Location
Chr.19: 43872363-43883964 on Build GRCh38
Transcripts
NM_001033719.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5714985 c.664A>G p.K222E Chr.19: 43873550-43873550 on Build GRCh38
5714986 c.658A>G p.K220E Chr.19: 43873550-43873550 on Build GRCh38
7126298 c.697C>T p.H233Y Chr.19: 43873517-43873517 on Build GRCh38
3289469 c.760C>T p.R254* Chr.19: 43873448-43873448 on Build GRCh38
3289473 c.630G>A p.M210I Chr.19: 43873578-43873578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76483217 Chr.19: 43873462-43873462 on Build GRCh38
rs749171632 Chr.19: 43873546-43873546 on Build GRCh38
rs376219255 Chr.19: 43873522-43873522 on Build GRCh38
rs373038530 Chr.19: 43873587-43873587 on Build GRCh38
rs527778126 Chr.19: 43873584-43873584 on Build GRCh38
rs35756172 Chr.19: 43873466-43873466 on Build GRCh38
rs750515051 Chr.19: 43873527-43873527 on Build GRCh38
rs144201116 Chr.19: 43873461-43873461 on Build GRCh38
rs769536602 Chr.19: 43873538-43873538 on Build GRCh38
rs12978958 Chr.19: 43873576-43873576 on Build GRCh38
rs760987927 Chr.19: 43873513-43873513 on Build GRCh38
rs370102344 Chr.19: 43873586-43873586 on Build GRCh38
rs753068493 Chr.19: 43873504-43873504 on Build GRCh38
rs192575521 Chr.19: 43873514-43873514 on Build GRCh38
rs369554464 Chr.19: 43873537-43873537 on Build GRCh38
rs759152735 Chr.19: 43873555-43873555 on Build GRCh38
rs548924610 Chr.19: 43873611-43873611 on Build GRCh38
rs764429040 Chr.19: 43873585-43873585 on Build GRCh38
rs560369991 Chr.19: 43873589-43873589 on Build GRCh38
rs759211072 Chr.19: 43873500-43873500 on Build GRCh38
rs79582952 Chr.19: 43873554-43873554 on Build GRCh38
rs758565954 Chr.19: 43873529-43873529 on Build GRCh38
rs12977303 Chr.19: 43873517-43873517 on Build GRCh38
rs751796267 Chr.19: 43873631-43873631 on Build GRCh38
rs1032376405 Chr.19: 43873481-43873481 on Build GRCh38
rs757321795 Chr.19: 43873519-43873519 on Build GRCh38
rs780170098 Chr.19: 43873530-43873530 on Build GRCh38
rs777763527 Chr.19: 43873542-43873542 on Build GRCh38
rs766612437 Chr.19: 43873628-43873628 on Build GRCh38
rs373749668 Chr.19: 43873553-43873553 on Build GRCh38
rs771728040 Chr.19: 43873565-43873565 on Build GRCh38
rs767101610 Chr.19: 43873503-43873503 on Build GRCh38
rs992069346 Chr.19: 43873477-43873477 on Build GRCh38
rs961371733 Chr.19: 43873545-43873545 on Build GRCh38
rs775057171 Chr.19: 43873572-43873572 on Build GRCh38
rs374757295 Chr.19: 43873488-43873488 on Build GRCh38
rs377424599 Chr.19: 43873605-43873605 on Build GRCh38
rs775686590 Chr.19: 43873582-43873595 on Build GRCh38

View Full Product Details