Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35550707-35550936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4A
Entrez Gene ID
495
Gene Name
ATPase H+/K+ transporting alpha subunit
Gene Aliases
ATP6A
UniGene
Hs.36992
Gene Chromosome Location
Chr.19: 35550193-35563658 on Build GRCh38
Transcripts
NM_000704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356990 c.3031T>A p.F1011I Chr.19: 35550882-35550882 on Build GRCh38
1718120 c.3050G>A p.R1017Q Chr.19: 35550863-35550863 on Build GRCh38
5626744 c.3033C>T p.F1011F Chr.19: 35550880-35550880 on Build GRCh38
6586307 c.3021C>T p.G1007G Chr.19: 35550892-35550892 on Build GRCh38
3938022 c.3065G>A p.R1022H Chr.19: 35550848-35550848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769534360 Chr.19: 35550803-35550803 on Build GRCh38
rs1042605251 Chr.19: 35550769-35550769 on Build GRCh38
rs772372492 Chr.19: 35550831-35550831 on Build GRCh38
rs146641851 Chr.19: 35550894-35550894 on Build GRCh38
rs763693283 Chr.19: 35550896-35550896 on Build GRCh38
rs760879648 Chr.19: 35550849-35550849 on Build GRCh38
rs775351403 Chr.19: 35550808-35550808 on Build GRCh38
rs1044505486 Chr.19: 35550729-35550729 on Build GRCh38
rs201163746 Chr.19: 35550785-35550785 on Build GRCh38
rs141390472 Chr.19: 35550895-35550895 on Build GRCh38
rs148503854 Chr.19: 35550879-35550879 on Build GRCh38
rs555560469 Chr.19: 35550772-35550772 on Build GRCh38
rs751179419 Chr.19: 35550904-35550904 on Build GRCh38
rs182728369 Chr.19: 35550778-35550778 on Build GRCh38
rs201486149 Chr.19: 35550791-35550791 on Build GRCh38
rs185924462 Chr.19: 35550856-35550856 on Build GRCh38
rs144537248 Chr.19: 35550883-35550883 on Build GRCh38
rs768722481 Chr.19: 35550810-35550810 on Build GRCh38
rs535319850 Chr.19: 35550745-35550745 on Build GRCh38
rs748332451 Chr.19: 35550814-35550814 on Build GRCh38
rs756807518 Chr.19: 35550908-35550908 on Build GRCh38
rs896980014 Chr.19: 35550880-35550880 on Build GRCh38
rs559698579 Chr.19: 35550906-35550906 on Build GRCh38
rs907457150 Chr.19: 35550741-35550741 on Build GRCh38
rs1046288378 Chr.19: 35550864-35550864 on Build GRCh38
rs1026923996 Chr.19: 35550886-35550886 on Build GRCh38
rs945655761 Chr.19: 35550767-35550767 on Build GRCh38
rs745794777 Chr.19: 35550792-35550792 on Build GRCh38
rs777066391 Chr.19: 35550875-35550875 on Build GRCh38
rs773459189 Chr.19: 35550848-35550848 on Build GRCh38
rs867856832 Chr.19: 35550782-35550782 on Build GRCh38
rs1003119328 Chr.19: 35550754-35550754 on Build GRCh38
rs755477472 Chr.19: 35550779-35550779 on Build GRCh38

View Full Product Details