Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38307605-38307858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A14
Entrez Gene ID
9389
Gene Name
solute carrier family 22 member 14
Gene Aliases
OCTL2, OCTL4, ORCTL4
UniGene
Hs.165559
Gene Chromosome Location
Chr.3: 38281662-38318575 on Build GRCh38
Transcripts
NM_001320033.1 NM_004803.4 XM_005265585.4 XM_006713416.3 XM_006713417.3 XM_006713418.3 XM_011534245.2 XM_011534247.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2986146 c.661C>A p.L221M Chr.3: 38307606-38307606 on Build GRCh38
6035962 c.753C>T p.Y251Y Chr.3: 38307698-38307698 on Build GRCh38
6780200 c.775+2T>C p.? Chr.3: 38307722-38307722 on Build GRCh38
3593194 c.673G>A p.G225S Chr.3: 38307618-38307618 on Build GRCh38
3593195 c.721C>T p.R241C Chr.3: 38307666-38307666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373067317 Chr.3: 38307770-38307772 on Build GRCh38
rs201320372 Chr.3: 38307723-38307723 on Build GRCh38
rs773018006 Chr.3: 38307734-38307734 on Build GRCh38
rs779826305 Chr.3: 38307629-38307629 on Build GRCh38
rs772666811 Chr.3: 38307746-38307746 on Build GRCh38
rs754801833 Chr.3: 38307726-38307726 on Build GRCh38
rs201709407 Chr.3: 38307737-38307737 on Build GRCh38
rs894890077 Chr.3: 38307815-38307815 on Build GRCh38
rs546996289 Chr.3: 38307666-38307666 on Build GRCh38
rs559918599 Chr.3: 38307767-38307769 on Build GRCh38
rs386660259 Chr.3: 38307675-38307676 on Build GRCh38
rs113828099 Chr.3: 38307736-38307736 on Build GRCh38
rs906335226 Chr.3: 38307777-38307777 on Build GRCh38
rs752026810 Chr.3: 38307686-38307686 on Build GRCh38
rs536419043 Chr.3: 38307808-38307808 on Build GRCh38
rs200149889 Chr.3: 38307699-38307699 on Build GRCh38
rs776239197 Chr.3: 38307655-38307655 on Build GRCh38
rs947567621 Chr.3: 38307791-38307791 on Build GRCh38
rs748815997 Chr.3: 38307747-38307747 on Build GRCh38
rs768349296 Chr.3: 38307651-38307651 on Build GRCh38
rs1078846 Chr.3: 38307679-38307679 on Build GRCh38
rs372653471 Chr.3: 38307694-38307694 on Build GRCh38
rs764091114 Chr.3: 38307714-38307714 on Build GRCh38
rs995017216 Chr.3: 38307834-38307834 on Build GRCh38
rs752893960 Chr.3: 38307695-38307695 on Build GRCh38
rs771999101 Chr.3: 38307682-38307682 on Build GRCh38
rs778654073 Chr.3: 38307750-38307750 on Build GRCh38
rs572026081 Chr.3: 38307656-38307657 on Build GRCh38
rs1003384234 Chr.3: 38307809-38307809 on Build GRCh38
rs146612743 Chr.3: 38307667-38307667 on Build GRCh38
rs746796831 Chr.3: 38307644-38307644 on Build GRCh38
rs760809449 Chr.3: 38307770-38307770 on Build GRCh38
rs542959928 Chr.3: 38307675-38307675 on Build GRCh38
rs969129103 Chr.3: 38307782-38307782 on Build GRCh38
rs9873449 Chr.3: 38307717-38307717 on Build GRCh38
rs750293511 Chr.3: 38307751-38307752 on Build GRCh38
rs758394182 Chr.3: 38307720-38307720 on Build GRCh38
rs780991631 Chr.3: 38307733-38307733 on Build GRCh38
rs973082896 Chr.3: 38307692-38307692 on Build GRCh38
rs148000898 Chr.3: 38307680-38307680 on Build GRCh38
rs759955793 Chr.3: 38307687-38307687 on Build GRCh38
rs182743293 Chr.3: 38307837-38307837 on Build GRCh38
rs1043325963 Chr.3: 38307813-38307813 on Build GRCh38
rs756971868 Chr.3: 38307695-38307695 on Build GRCh38
rs556532794 Chr.3: 38307676-38307676 on Build GRCh38
rs138257211 Chr.3: 38307656-38307659 on Build GRCh38
rs374016281 Chr.3: 38307704-38307704 on Build GRCh38
rs190905770 Chr.3: 38307814-38307814 on Build GRCh38
rs758128590 Chr.3: 38307759-38307760 on Build GRCh38
rs867504422 Chr.3: 38307810-38307810 on Build GRCh38
rs1027770575 Chr.3: 38307705-38307705 on Build GRCh38
rs769423383 Chr.3: 38307738-38307738 on Build GRCh38
rs141638661 Chr.3: 38307769-38307769 on Build GRCh38
rs977203916 Chr.3: 38307743-38307743 on Build GRCh38
rs371887827 Chr.3: 38307763-38307763 on Build GRCh38
rs199741711 Chr.3: 38307754-38307754 on Build GRCh38
rs774967688 Chr.3: 38307761-38307761 on Build GRCh38
rs201212757 Chr.3: 38307768-38307768 on Build GRCh38

View Full Product Details