Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100665062-100665311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRPX2
Entrez Gene ID
27286
Gene Name
sushi repeat containing protein, X-linked 2
Gene Aliases
BPP, CBPS, PMGX, RESDX, SRPUL
UniGene
Hs.306339
Gene Chromosome Location
Chr.X: 100644166-100671299 on Build GRCh38
Transcripts
NM_014467.2

SNP Detail

SNP ID Chromosome Location
rs962218701 Chr.X: 100665228-100665228 on Build GRCh38
rs755581646 Chr.X: 100665212-100665212 on Build GRCh38
rs771790252 Chr.X: 100665278-100665278 on Build GRCh38
rs1053428544 Chr.X: 100665189-100665189 on Build GRCh38
rs779500463 Chr.X: 100665256-100665256 on Build GRCh38
rs1009471156 Chr.X: 100665208-100665208 on Build GRCh38
rs368046912 Chr.X: 100665205-100665205 on Build GRCh38
rs370080258 Chr.X: 100665263-100665263 on Build GRCh38
rs181200947 Chr.X: 100665126-100665126 on Build GRCh38
rs993778885 Chr.X: 100665247-100665247 on Build GRCh38
rs149317696 Chr.X: 100665274-100665274 on Build GRCh38
rs745455864 Chr.X: 100665249-100665249 on Build GRCh38
rs146282439 Chr.X: 100665279-100665279 on Build GRCh38
rs943132618 Chr.X: 100665141-100665141 on Build GRCh38
rs757793078 Chr.X: 100665248-100665248 on Build GRCh38
rs759091523 Chr.X: 100665111-100665111 on Build GRCh38
rs910264449 Chr.X: 100665132-100665132 on Build GRCh38
rs758792281 Chr.X: 100665233-100665233 on Build GRCh38
rs768629864 Chr.X: 100665264-100665264 on Build GRCh38
rs185564106 Chr.X: 100665213-100665213 on Build GRCh38
rs890871264 Chr.X: 100665128-100665128 on Build GRCh38
rs796053343 Chr.X: 100665242-100665242 on Build GRCh38
rs769478434 Chr.X: 100665252-100665252 on Build GRCh38

View Full Product Details