Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30379362-30379611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT1
Entrez Gene ID
1731
Gene Name
septin 1
Gene Aliases
DIFF6, LARP, PNUTL3, SEP1
UniGene
Hs.632176
Gene Chromosome Location
Chr.16: 30378133-30382850 on Build GRCh38
Transcripts
NM_052838.4 XM_017022991.1 NM_001214906.1 NM_001214907.1

Gene Symbol
ZNF48
Entrez Gene ID
197407
Gene Name
zinc finger protein 48
Gene Aliases
ZNF553
UniGene
Hs.513501
Gene Chromosome Location
Chr.16: 30378312-30400108 on Build GRCh38
Transcripts
NM_052838.4 XM_017022991.1 NM_001214906.1 NM_001214907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1225224 c.737G>A p.R246H Chr.16: 30379458-30379458 on Build GRCh38
3948472 c.729G>C p.R243S Chr.16: 30379466-30379466 on Build GRCh38
3377866 c.753C>A p.T251T Chr.16: 30379442-30379442 on Build GRCh38
4824693 c.714G>A p.G238G Chr.16: 30379481-30379481 on Build GRCh38
703297 c.661G>A p.E221K Chr.16: 30379534-30379534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755635579 Chr.16: 30379533-30379533 on Build GRCh38
rs775771282 Chr.16: 30379551-30379551 on Build GRCh38
rs747393569 Chr.16: 30379554-30379554 on Build GRCh38
rs761914888 Chr.16: 30379579-30379579 on Build GRCh38
rs886213920 Chr.16: 30379394-30379394 on Build GRCh38
rs750235408 Chr.16: 30379389-30379389 on Build GRCh38
rs769576285 Chr.16: 30379414-30379414 on Build GRCh38
rs777598329 Chr.16: 30379539-30379539 on Build GRCh38
rs765469218 Chr.16: 30379477-30379477 on Build GRCh38
rs754938047 Chr.16: 30379501-30379501 on Build GRCh38
rs975811089 Chr.16: 30379395-30379395 on Build GRCh38
rs747625480 Chr.16: 30379512-30379512 on Build GRCh38
rs978544981 Chr.16: 30379498-30379498 on Build GRCh38
rs371586167 Chr.16: 30379499-30379499 on Build GRCh38
rs367740501 Chr.16: 30379577-30379577 on Build GRCh38
rs780625170 Chr.16: 30379403-30379403 on Build GRCh38
rs754922578 Chr.16: 30379402-30379402 on Build GRCh38
rs368210646 Chr.16: 30379397-30379397 on Build GRCh38
rs761882617 Chr.16: 30379467-30379467 on Build GRCh38
rs764645069 Chr.16: 30379460-30379460 on Build GRCh38
rs368140022 Chr.16: 30379563-30379563 on Build GRCh38
rs758143945 Chr.16: 30379392-30379392 on Build GRCh38
rs374254859 Chr.16: 30379387-30379387 on Build GRCh38
rs984495471 Chr.16: 30379573-30379573 on Build GRCh38
rs758591486 Chr.16: 30379490-30379490 on Build GRCh38
rs540193596 Chr.16: 30379588-30379588 on Build GRCh38
rs772941508 Chr.16: 30379442-30379442 on Build GRCh38
rs770682010 Chr.16: 30379548-30379548 on Build GRCh38
rs960799078 Chr.16: 30379513-30379513 on Build GRCh38
rs781120091 Chr.16: 30379502-30379502 on Build GRCh38
rs775891343 Chr.16: 30379458-30379458 on Build GRCh38
rs554529920 Chr.16: 30379583-30379583 on Build GRCh38
rs780935716 Chr.16: 30379449-30379450 on Build GRCh38
rs374177132 Chr.16: 30379462-30379462 on Build GRCh38
rs137903946 Chr.16: 30379497-30379497 on Build GRCh38
rs371117855 Chr.16: 30379459-30379459 on Build GRCh38
rs746963085 Chr.16: 30379399-30379399 on Build GRCh38
rs998452872 Chr.16: 30379441-30379441 on Build GRCh38
rs750606776 Chr.16: 30379484-30379484 on Build GRCh38
rs772524373 Chr.16: 30379451-30379451 on Build GRCh38
rs747691315 Chr.16: 30379412-30379412 on Build GRCh38
rs748987262 Chr.16: 30379547-30379547 on Build GRCh38

View Full Product Details