Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56621514-56621769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF71
Entrez Gene ID
58491
Gene Name
zinc finger protein 71
Gene Aliases
EZFIT
UniGene
Hs.301431
Gene Chromosome Location
Chr.19: 56595259-56624481 on Build GRCh38
Transcripts
NM_021216.4 XM_011527193.2 XM_017027078.1 XM_017027079.1 XM_017027080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1001937 c.305G>A p.C102Y Chr.19: 56621592-56621592 on Build GRCh38
4081856 c.322G>A p.G108S Chr.19: 56621609-56621609 on Build GRCh38
4081857 c.323G>A p.G108D Chr.19: 56621610-56621610 on Build GRCh38
6482002 c.234G>A p.L78L Chr.19: 56621521-56621521 on Build GRCh38
1001938 c.444C>T p.H148H Chr.19: 56621731-56621731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745370886 Chr.19: 56621717-56621717 on Build GRCh38
rs756225557 Chr.19: 56621686-56621686 on Build GRCh38
rs142626174 Chr.19: 56621675-56621675 on Build GRCh38
rs1053730850 Chr.19: 56621646-56621646 on Build GRCh38
rs139335499 Chr.19: 56621607-56621607 on Build GRCh38
rs746862574 Chr.19: 56621618-56621618 on Build GRCh38
rs752789246 Chr.19: 56621660-56621662 on Build GRCh38
rs575206556 Chr.19: 56621677-56621677 on Build GRCh38
rs142544600 Chr.19: 56621574-56621574 on Build GRCh38
rs774093863 Chr.19: 56621558-56621558 on Build GRCh38
rs373781920 Chr.19: 56621605-56621605 on Build GRCh38
rs770904153 Chr.19: 56621622-56621622 on Build GRCh38
rs746022630 Chr.19: 56621549-56621549 on Build GRCh38
rs35392779 Chr.19: 56621649-56621649 on Build GRCh38
rs896350281 Chr.19: 56621642-56621642 on Build GRCh38
rs368745207 Chr.19: 56621716-56621716 on Build GRCh38
rs773062752 Chr.19: 56621562-56621562 on Build GRCh38
rs776216179 Chr.19: 56621582-56621582 on Build GRCh38
rs200965626 Chr.19: 56621599-56621599 on Build GRCh38
rs367626222 Chr.19: 56621606-56621606 on Build GRCh38
rs374454585 Chr.19: 56621746-56621746 on Build GRCh38
rs762189547 Chr.19: 56621676-56621676 on Build GRCh38
rs2072501 Chr.19: 56621600-56621600 on Build GRCh38
rs759189672 Chr.19: 56621585-56621585 on Build GRCh38
rs781068556 Chr.19: 56621604-56621604 on Build GRCh38
rs770721727 Chr.19: 56621715-56621715 on Build GRCh38
rs1056682615 Chr.19: 56621626-56621626 on Build GRCh38
rs377218850 Chr.19: 56621550-56621550 on Build GRCh38
rs777177328 Chr.19: 56621543-56621544 on Build GRCh38
rs762950166 Chr.19: 56621736-56621745 on Build GRCh38
rs371046743 Chr.19: 56621714-56621714 on Build GRCh38
rs994447919 Chr.19: 56621742-56621742 on Build GRCh38
rs769489986 Chr.19: 56621739-56621739 on Build GRCh38
rs1044862120 Chr.19: 56621656-56621656 on Build GRCh38
rs760226929 Chr.19: 56621570-56621570 on Build GRCh38
rs146422242 Chr.19: 56621709-56621709 on Build GRCh38
rs781168570 Chr.19: 56621644-56621645 on Build GRCh38
rs755239629 Chr.19: 56621692-56621692 on Build GRCh38
rs1029852938 Chr.19: 56621555-56621555 on Build GRCh38
rs147756631 Chr.19: 56621660-56621660 on Build GRCh38
rs750480173 Chr.19: 56621678-56621678 on Build GRCh38
rs780249877 Chr.19: 56621687-56621687 on Build GRCh38
rs199651640 Chr.19: 56621587-56621587 on Build GRCh38
rs148434207 Chr.19: 56621569-56621569 on Build GRCh38
rs777751379 Chr.19: 56621610-56621610 on Build GRCh38
rs372176440 Chr.19: 56621556-56621556 on Build GRCh38
rs758495622 Chr.19: 56621635-56621635 on Build GRCh38
rs775354467 Chr.19: 56621650-56621650 on Build GRCh38
rs754116323 Chr.19: 56621691-56621691 on Build GRCh38
rs536091468 Chr.19: 56621541-56621541 on Build GRCh38
rs764661500 Chr.19: 56621596-56621596 on Build GRCh38
rs753057359 Chr.19: 56621588-56621588 on Build GRCh38
rs200440090 Chr.19: 56621623-56621623 on Build GRCh38
rs370465514 Chr.19: 56621559-56621559 on Build GRCh38
rs186242664 Chr.19: 56621674-56621674 on Build GRCh38
rs775961190 Chr.19: 56621648-56621648 on Build GRCh38
rs763455008 Chr.19: 56621589-56621589 on Build GRCh38
rs765730216 Chr.19: 56621561-56621561 on Build GRCh38
rs760100841 Chr.19: 56621579-56621580 on Build GRCh38
rs747899140 Chr.19: 56621745-56621745 on Build GRCh38

View Full Product Details