Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28842984-28843231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUFM
Entrez Gene ID
7284
Gene Name
Tu translation elongation factor, mitochondrial
Gene Aliases
COXPD4, EF-TuMT, EFTU, P43
UniGene
Hs.12084
Gene Chromosome Location
Chr.16: 28842411-28859562 on Build GRCh38
Transcripts
NM_003321.4 XM_011545928.2 XM_017023619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5788572 c.1312C>T p.L438L Chr.16: 28843031-28843031 on Build GRCh38
6406284 c.1345G>T p.E449* Chr.16: 28842998-28842998 on Build GRCh38
7270201 c.1325C>T p.T442M Chr.16: 28843018-28843018 on Build GRCh38
6505754 c.1288G>A p.G430S Chr.16: 28843055-28843055 on Build GRCh38
375001 c.1308C>T p.T436T Chr.16: 28843035-28843035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749592250 Chr.16: 28843134-28843134 on Build GRCh38
rs756481272 Chr.16: 28843043-28843043 on Build GRCh38
rs746833692 Chr.16: 28843087-28843087 on Build GRCh38
rs898638015 Chr.16: 28843044-28843044 on Build GRCh38
rs753214812 Chr.16: 28843102-28843102 on Build GRCh38
rs143510655 Chr.16: 28843094-28843094 on Build GRCh38
rs199806073 Chr.16: 28843101-28843101 on Build GRCh38
rs753588078 Chr.16: 28843131-28843131 on Build GRCh38
rs146326033 Chr.16: 28843051-28843051 on Build GRCh38
rs756571326 Chr.16: 28843114-28843114 on Build GRCh38
rs756857990 Chr.16: 28843133-28843133 on Build GRCh38
rs779968922 Chr.16: 28843128-28843128 on Build GRCh38
rs546848930 Chr.16: 28843202-28843203 on Build GRCh38
rs376169369 Chr.16: 28843151-28843151 on Build GRCh38
rs863224246 Chr.16: 28843147-28843147 on Build GRCh38
rs1024963469 Chr.16: 28843113-28843113 on Build GRCh38
rs767587350 Chr.16: 28843183-28843183 on Build GRCh38
rs753124586 Chr.16: 28843035-28843035 on Build GRCh38
rs752146538 Chr.16: 28843017-28843017 on Build GRCh38
rs749568845 Chr.16: 28843164-28843164 on Build GRCh38
rs117230775 Chr.16: 28843119-28843119 on Build GRCh38
rs139550011 Chr.16: 28843174-28843174 on Build GRCh38
rs781663104 Chr.16: 28843033-28843033 on Build GRCh38
rs770526342 Chr.16: 28843052-28843052 on Build GRCh38
rs779985614 Chr.16: 28843158-28843158 on Build GRCh38
rs777602400 Chr.16: 28843050-28843050 on Build GRCh38
rs529976762 Chr.16: 28843091-28843091 on Build GRCh38
rs745888993 Chr.16: 28843067-28843067 on Build GRCh38
rs769966138 Chr.16: 28843120-28843120 on Build GRCh38
rs771893374 Chr.16: 28843072-28843072 on Build GRCh38
rs759602167 Chr.16: 28843095-28843095 on Build GRCh38
rs771071469 Chr.16: 28843165-28843165 on Build GRCh38
rs778258598 Chr.16: 28843054-28843054 on Build GRCh38
rs745387851 Chr.16: 28843139-28843139 on Build GRCh38
rs775427401 Chr.16: 28843073-28843073 on Build GRCh38
rs768388341 Chr.16: 28843089-28843089 on Build GRCh38
rs369357902 Chr.16: 28843161-28843161 on Build GRCh38
rs768476195 Chr.16: 28843160-28843160 on Build GRCh38
rs145028779 Chr.16: 28843018-28843018 on Build GRCh38
rs990653187 Chr.16: 28843202-28843202 on Build GRCh38
rs776189219 Chr.16: 28843199-28843199 on Build GRCh38
rs759567579 Chr.16: 28843175-28843175 on Build GRCh38

View Full Product Details