Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7549415-7549656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF12
Entrez Gene ID
8742
Gene Name
tumor necrosis factor superfamily member 12
Gene Aliases
APO3L, DR3LG, TNLG4A, TWEAK
UniGene
Hs.54673
Gene Chromosome Location
Chr.17: 7549058-7557890 on Build GRCh38
Transcripts
NM_003809.2 NR_037146.1 NM_172089.3

Gene Symbol
TNFSF12-TNFSF13
Entrez Gene ID
407977
Gene Name
TNFSF12-TNFSF13 readthrough
Gene Aliases
TWE-PRIL
UniGene
Hs.54673
Gene Chromosome Location
Chr.17: 7549058-7561608 on Build GRCh38
Transcripts
NM_003809.2 NR_037146.1 NM_172089.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
215718 c.202C>G p.P68A Chr.17: 7549516-7549516 on Build GRCh38
6465183 c.168C>T p.A56A Chr.17: 7549482-7549482 on Build GRCh38
215717 c.202C>G p.P68A Chr.17: 7549516-7549516 on Build GRCh38
6465182 c.168C>T p.A56A Chr.17: 7549482-7549482 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868687352 Chr.17: 7549491-7549491 on Build GRCh38
rs376316318 Chr.17: 7549579-7549579 on Build GRCh38
rs552469927 Chr.17: 7549464-7549464 on Build GRCh38
rs938723059 Chr.17: 7549596-7549596 on Build GRCh38
rs778908741 Chr.17: 7549552-7549552 on Build GRCh38
rs781196267 Chr.17: 7549531-7549531 on Build GRCh38
rs764682956 Chr.17: 7549459-7549459 on Build GRCh38
rs867531939 Chr.17: 7549481-7549481 on Build GRCh38
rs571042380 Chr.17: 7549465-7549465 on Build GRCh38
rs754711946 Chr.17: 7549520-7549520 on Build GRCh38
rs894641102 Chr.17: 7549609-7549609 on Build GRCh38
rs149475179 Chr.17: 7549460-7549460 on Build GRCh38
rs772401999 Chr.17: 7549441-7549441 on Build GRCh38
rs747019973 Chr.17: 7549508-7549508 on Build GRCh38
rs751381442 Chr.17: 7549518-7549518 on Build GRCh38
rs137994694 Chr.17: 7549455-7549455 on Build GRCh38
rs1057269782 Chr.17: 7549597-7549597 on Build GRCh38
rs749918168 Chr.17: 7549488-7549488 on Build GRCh38
rs764718533 Chr.17: 7549478-7549478 on Build GRCh38
rs753259588 Chr.17: 7549516-7549517 on Build GRCh38
rs908631855 Chr.17: 7549594-7549594 on Build GRCh38
rs776037378 Chr.17: 7549454-7549454 on Build GRCh38
rs772095320 Chr.17: 7549607-7549607 on Build GRCh38
rs767853982 Chr.17: 7549435-7549454 on Build GRCh38
rs866471555 Chr.17: 7549486-7549486 on Build GRCh38
rs74578413 Chr.17: 7549589-7549589 on Build GRCh38
rs779644920 Chr.17: 7549517-7549517 on Build GRCh38
rs1043271702 Chr.17: 7549626-7549626 on Build GRCh38
rs758090917 Chr.17: 7549504-7549504 on Build GRCh38
rs80067372 Chr.17: 7549435-7549435 on Build GRCh38
rs756221603 Chr.17: 7549561-7549563 on Build GRCh38
rs769757755 Chr.17: 7549539-7549539 on Build GRCh38
rs867626890 Chr.17: 7549560-7549560 on Build GRCh38
rs775735044 Chr.17: 7549610-7549610 on Build GRCh38
rs778020698 Chr.17: 7549468-7549468 on Build GRCh38
rs534969361 Chr.17: 7549538-7549538 on Build GRCh38

View Full Product Details