Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137008750-137008997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA2
Entrez Gene ID
20
Gene Name
ATP binding cassette subfamily A member 2
Gene Aliases
ABC2
UniGene
Hs.421202
Gene Chromosome Location
Chr.9: 137007234-137028922 on Build GRCh38
Transcripts
NM_001606.4 NM_212533.2 XM_006716996.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905912 c.6885C>T p.D2295D Chr.9: 137008996-137008996 on Build GRCh38
6115357 c.6886G>T p.V2296L Chr.9: 137008995-137008995 on Build GRCh38
6589640 c.6935G>A p.R2312Q Chr.9: 137008864-137008864 on Build GRCh38
5410780 c.6939C>T p.H2313H Chr.9: 137008860-137008860 on Build GRCh38
5965251 c.6974A>G p.H2325R Chr.9: 137008825-137008825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146707252 Chr.9: 137008939-137008939 on Build GRCh38
rs550071486 Chr.9: 137008954-137008954 on Build GRCh38
rs774493213 Chr.9: 137008806-137008806 on Build GRCh38
rs763492187 Chr.9: 137008830-137008830 on Build GRCh38
rs543664782 Chr.9: 137008857-137008857 on Build GRCh38
rs745794491 Chr.9: 137008917-137008917 on Build GRCh38
rs755662629 Chr.9: 137008873-137008873 on Build GRCh38
rs200606263 Chr.9: 137008944-137008944 on Build GRCh38
rs371871913 Chr.9: 137008913-137008913 on Build GRCh38
rs780108634 Chr.9: 137008979-137008979 on Build GRCh38
rs575923299 Chr.9: 137008871-137008871 on Build GRCh38
rs765822195 Chr.9: 137008946-137008946 on Build GRCh38
rs769729071 Chr.9: 137008918-137008918 on Build GRCh38
rs775730867 Chr.9: 137008904-137008905 on Build GRCh38
rs767037178 Chr.9: 137008826-137008826 on Build GRCh38
rs753298020 Chr.9: 137008899-137008900 on Build GRCh38
rs771891462 Chr.9: 137008901-137008902 on Build GRCh38
rs755448053 Chr.9: 137008894-137008894 on Build GRCh38
rs368650540 Chr.9: 137008928-137008928 on Build GRCh38
rs201625278 Chr.9: 137008842-137008842 on Build GRCh38
rs751226065 Chr.9: 137008956-137008956 on Build GRCh38
rs779572218 Chr.9: 137008874-137008874 on Build GRCh38
rs397761371 Chr.9: 137008907-137008908 on Build GRCh38
rs375127084 Chr.9: 137008907-137008907 on Build GRCh38
rs748302984 Chr.9: 137008938-137008938 on Build GRCh38
rs762034018 Chr.9: 137008808-137008808 on Build GRCh38
rs375671010 Chr.9: 137008915-137008915 on Build GRCh38
rs371695208 Chr.9: 137008976-137008976 on Build GRCh38
rs750062304 Chr.9: 137008848-137008850 on Build GRCh38
rs773629012 Chr.9: 137008941-137008941 on Build GRCh38
rs758029250 Chr.9: 137008856-137008856 on Build GRCh38
rs755839713 Chr.9: 137008977-137008977 on Build GRCh38
rs866545067 Chr.9: 137008900-137008900 on Build GRCh38
rs761192828 Chr.9: 137008878-137008878 on Build GRCh38
rs759982951 Chr.9: 137008945-137008945 on Build GRCh38
rs79419594 Chr.9: 137008875-137008875 on Build GRCh38
rs754412530 Chr.9: 137008892-137008892 on Build GRCh38
rs753112833 Chr.9: 137008947-137008947 on Build GRCh38
rs201682816 Chr.9: 137008818-137008818 on Build GRCh38
rs781618092 Chr.9: 137008902-137008903 on Build GRCh38
rs776613952 Chr.9: 137008874-137008874 on Build GRCh38
rs763059738 Chr.9: 137008784-137008784 on Build GRCh38
rs199815377 Chr.9: 137008864-137008864 on Build GRCh38
rs754394147 Chr.9: 137008832-137008832 on Build GRCh38
rs201340591 Chr.9: 137008943-137008943 on Build GRCh38
rs756832150 Chr.9: 137008966-137008966 on Build GRCh38
rs770097467 Chr.9: 137008903-137008904 on Build GRCh38
rs764686232 Chr.9: 137008899-137008899 on Build GRCh38
rs770807217 Chr.9: 137008778-137008778 on Build GRCh38
rs754507796 Chr.9: 137008836-137008836 on Build GRCh38
rs747761832 Chr.9: 137008848-137008848 on Build GRCh38
rs766840634 Chr.9: 137008885-137008885 on Build GRCh38
rs59031144 Chr.9: 137008900-137008901 on Build GRCh38
rs760924667 Chr.9: 137008942-137008942 on Build GRCh38
rs756528457 Chr.9: 137008824-137008824 on Build GRCh38
rs954868910 Chr.9: 137008797-137008797 on Build GRCh38
rs773541051 Chr.9: 137008877-137008877 on Build GRCh38
rs764228129 Chr.9: 137008800-137008800 on Build GRCh38
rs747962687 Chr.9: 137008901-137008901 on Build GRCh38
rs527372692 Chr.9: 137008908-137008908 on Build GRCh38
rs561668236 Chr.9: 137008876-137008876 on Build GRCh38
rs763029236 Chr.9: 137008906-137008907 on Build GRCh38
rs780992044 Chr.9: 137008967-137008967 on Build GRCh38
rs370558699 Chr.9: 137008779-137008779 on Build GRCh38
rs368283479 Chr.9: 137008912-137008912 on Build GRCh38

View Full Product Details