Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96852646-96852907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VRK1
Entrez Gene ID
7443
Gene Name
vaccinia related kinase 1
Gene Aliases
PCH1, PCH1A
UniGene
Hs.422662
Gene Chromosome Location
Chr.14: 96797309-96881614 on Build GRCh38
Transcripts
NM_003384.2 XM_006720247.3 XM_011537132.1 XM_017021622.1 XM_017021624.1 XM_017021625.1 XM_017021626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
959433 c.398G>A p.R133H Chr.14: 96852854-96852854 on Build GRCh38
4492436 c.397C>T p.R133C Chr.14: 96852853-96852853 on Build GRCh38
3499618 c.451C>T p.R151W Chr.14: 96852907-96852907 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764788560 Chr.14: 96852874-96852874 on Build GRCh38
rs915524591 Chr.14: 96852746-96852746 on Build GRCh38
rs186980733 Chr.14: 96852809-96852809 on Build GRCh38
rs758978677 Chr.14: 96852854-96852854 on Build GRCh38
rs387906830 Chr.14: 96852853-96852853 on Build GRCh38
rs370891173 Chr.14: 96852792-96852792 on Build GRCh38
rs770594833 Chr.14: 96852825-96852825 on Build GRCh38
rs778730789 Chr.14: 96852813-96852813 on Build GRCh38
rs764510402 Chr.14: 96852786-96852786 on Build GRCh38
rs551029072 Chr.14: 96852817-96852817 on Build GRCh38
rs756150767 Chr.14: 96852812-96852812 on Build GRCh38
rs772961519 Chr.14: 96852819-96852819 on Build GRCh38
rs753186957 Chr.14: 96852780-96852780 on Build GRCh38
rs533211396 Chr.14: 96852730-96852730 on Build GRCh38
rs368276508 Chr.14: 96852782-96852782 on Build GRCh38
rs7492598 Chr.14: 96852722-96852722 on Build GRCh38
rs191021502 Chr.14: 96852823-96852823 on Build GRCh38
rs375375557 Chr.14: 96852705-96852705 on Build GRCh38
rs745973904 Chr.14: 96852810-96852810 on Build GRCh38
rs946965871 Chr.14: 96852754-96852754 on Build GRCh38

View Full Product Details