Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 78697318-78697581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7
Entrez Gene ID
3574
Gene Name
interleukin 7
Gene Aliases
IL-7
UniGene
Hs.591873
Gene Chromosome Location
Chr.8: 78675870-78805523 on Build GRCh38
Transcripts
XM_011517523.2 NM_016010.2 XM_005251256.1 XM_006716454.2 XM_011517539.2

Gene Symbol
ZC2HC1A
Entrez Gene ID
51101
Gene Name
zinc finger C2HC-type containing 1A
Gene Aliases
C8orf70, CGI-62, FAM164A
UniGene
Hs.271876
Gene Chromosome Location
Chr.8: 78666047-78719765 on Build GRCh38
Transcripts
XM_011517523.2 NM_016010.2 XM_005251256.1 XM_006716454.2 XM_011517539.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4650092 c.566G>A p.R189Q Chr.8: 78697468-78697468 on Build GRCh38
6825544 c.517G>A p.A173T Chr.8: 78697419-78697419 on Build GRCh38
3901684 c.569T>C p.L190S Chr.8: 78697471-78697471 on Build GRCh38
1206172 c.565C>T p.R189* Chr.8: 78697467-78697467 on Build GRCh38
244370 c.537T>G p.S179S Chr.8: 78697439-78697439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764490342 Chr.8: 78697360-78697360 on Build GRCh38
rs1024344667 Chr.8: 78697408-78697408 on Build GRCh38
rs770560148 Chr.8: 78697433-78697433 on Build GRCh38
rs759250877 Chr.8: 78697387-78697387 on Build GRCh38
rs748867265 Chr.8: 78697424-78697424 on Build GRCh38
rs944859318 Chr.8: 78697489-78697489 on Build GRCh38
rs776838149 Chr.8: 78697434-78697434 on Build GRCh38
rs750516008 Chr.8: 78697475-78697475 on Build GRCh38
rs201671046 Chr.8: 78697488-78697488 on Build GRCh38
rs371962432 Chr.8: 78697468-78697468 on Build GRCh38
rs867047499 Chr.8: 78697492-78697492 on Build GRCh38
rs376457456 Chr.8: 78697523-78697523 on Build GRCh38
rs936698882 Chr.8: 78697449-78697449 on Build GRCh38
rs558401508 Chr.8: 78697394-78697394 on Build GRCh38
rs914163193 Chr.8: 78697486-78697486 on Build GRCh38
rs776941860 Chr.8: 78697446-78697446 on Build GRCh38
rs930890359 Chr.8: 78697431-78697431 on Build GRCh38
rs763173071 Chr.8: 78697487-78697487 on Build GRCh38
rs35904168 Chr.8: 78697412-78697412 on Build GRCh38
rs117424234 Chr.8: 78697518-78697518 on Build GRCh38
rs763422890 Chr.8: 78697549-78697549 on Build GRCh38
rs191233440 Chr.8: 78697348-78697348 on Build GRCh38
rs138000597 Chr.8: 78697419-78697419 on Build GRCh38
rs761746283 Chr.8: 78697378-78697378 on Build GRCh38
rs770062173 Chr.8: 78697510-78697510 on Build GRCh38
rs910678174 Chr.8: 78697460-78697460 on Build GRCh38
rs201882613 Chr.8: 78697358-78697358 on Build GRCh38
rs372858379 Chr.8: 78697418-78697418 on Build GRCh38
rs751288773 Chr.8: 78697417-78697417 on Build GRCh38
rs769129669 Chr.8: 78697421-78697421 on Build GRCh38
rs150344275 Chr.8: 78697414-78697414 on Build GRCh38
rs778393767 Chr.8: 78697546-78697546 on Build GRCh38
rs781044570 Chr.8: 78697500-78697500 on Build GRCh38
rs769083996 Chr.8: 78697440-78697440 on Build GRCh38
rs371705556 Chr.8: 78697519-78697519 on Build GRCh38
rs201338405 Chr.8: 78697527-78697527 on Build GRCh38
rs761256249 Chr.8: 78697497-78697497 on Build GRCh38
rs368633779 Chr.8: 78697423-78697423 on Build GRCh38
rs912619962 Chr.8: 78697349-78697349 on Build GRCh38
rs1055587327 Chr.8: 78697513-78697513 on Build GRCh38
rs371415546 Chr.8: 78697386-78697386 on Build GRCh38
rs754396802 Chr.8: 78697374-78697374 on Build GRCh38
rs765908988 Chr.8: 78697474-78697474 on Build GRCh38
rs745543222 Chr.8: 78697547-78697547 on Build GRCh38
rs780479715 Chr.8: 78697415-78697415 on Build GRCh38
rs200096220 Chr.8: 78697437-78697437 on Build GRCh38
rs765220994 Chr.8: 78697450-78697452 on Build GRCh38
rs777043052 Chr.8: 78697555-78697555 on Build GRCh38
rs951389788 Chr.8: 78697540-78697540 on Build GRCh38
rs377605327 Chr.8: 78697526-78697526 on Build GRCh38
rs906913023 Chr.8: 78697507-78697507 on Build GRCh38
rs760921799 Chr.8: 78697438-78697438 on Build GRCh38

View Full Product Details