Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52614176-52614437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF83
Entrez Gene ID
55769
Gene Name
zinc finger protein 83
Gene Aliases
HPF1, ZNF816B
UniGene
Hs.467210
Gene Chromosome Location
Chr.19: 52612364-52690581 on Build GRCh38
Transcripts
NM_001105549.1 NM_001105550.1 NM_001105551.1 NM_001105552.1 NM_001277945.1 NM_001277946.1 NM_001277947.1 NM_001277948.1 NM_001277949.1 NM_001277951.1 NM_001277952.1 NM_018300.3 XM_017026951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287053 c.252G>T p.E84D Chr.19: 52614313-52614313 on Build GRCh38
5622579 c.258A>G p.A86A Chr.19: 52614307-52614307 on Build GRCh38
6833479 c.179C>A p.S60Y Chr.19: 52614386-52614386 on Build GRCh38
5496758 c.162C>T p.S54S Chr.19: 52614403-52614403 on Build GRCh38
3938259 c.302C>A p.A101D Chr.19: 52614263-52614263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756878414 Chr.19: 52614245-52614245 on Build GRCh38
rs779696119 Chr.19: 52614270-52614270 on Build GRCh38
rs774190724 Chr.19: 52614380-52614381 on Build GRCh38
rs774686513 Chr.19: 52614393-52614393 on Build GRCh38
rs770264884 Chr.19: 52614316-52614316 on Build GRCh38
rs999137064 Chr.19: 52614309-52614309 on Build GRCh38
rs879147548 Chr.19: 52614404-52614404 on Build GRCh38
rs1056185 Chr.19: 52614278-52614278 on Build GRCh38
rs566790486 Chr.19: 52614392-52614392 on Build GRCh38
rs753602150 Chr.19: 52614310-52614310 on Build GRCh38
rs764039163 Chr.19: 52614308-52614308 on Build GRCh38
rs772219495 Chr.19: 52614399-52614399 on Build GRCh38
rs751614189 Chr.19: 52614216-52614216 on Build GRCh38
rs3786490 Chr.19: 52614356-52614356 on Build GRCh38
rs767555845 Chr.19: 52614220-52614220 on Build GRCh38
rs534154104 Chr.19: 52614394-52614394 on Build GRCh38
rs774598236 Chr.19: 52614286-52614286 on Build GRCh38
rs137930387 Chr.19: 52614235-52614235 on Build GRCh38
rs1030568220 Chr.19: 52614315-52614315 on Build GRCh38
rs756601169 Chr.19: 52614379-52614379 on Build GRCh38
rs750156258 Chr.19: 52614335-52614335 on Build GRCh38
rs778584351 Chr.19: 52614247-52614247 on Build GRCh38
rs774411419 Chr.19: 52614237-52614237 on Build GRCh38
rs778304085 Chr.19: 52614381-52614381 on Build GRCh38
rs549530184 Chr.19: 52614323-52614323 on Build GRCh38
rs757112901 Chr.19: 52614321-52614321 on Build GRCh38
rs567896850 Chr.19: 52614339-52614339 on Build GRCh38
rs77404321 Chr.19: 52614208-52614208 on Build GRCh38
rs367618930 Chr.19: 52614273-52614273 on Build GRCh38
rs749729407 Chr.19: 52614386-52614386 on Build GRCh38
rs329940 Chr.19: 52614209-52614209 on Build GRCh38
rs758028377 Chr.19: 52614249-52614249 on Build GRCh38
rs746243529 Chr.19: 52614291-52614291 on Build GRCh38
rs373776078 Chr.19: 52614238-52614238 on Build GRCh38
rs201376158 Chr.19: 52614352-52614352 on Build GRCh38
rs753606976 Chr.19: 52614244-52614244 on Build GRCh38
rs376657241 Chr.19: 52614288-52614288 on Build GRCh38
rs771090046 Chr.19: 52614283-52614283 on Build GRCh38
rs773462019 Chr.19: 52614276-52614276 on Build GRCh38
rs775547102 Chr.19: 52614292-52614292 on Build GRCh38
rs767643904 Chr.19: 52614289-52614289 on Build GRCh38
rs141660131 Chr.19: 52614272-52614272 on Build GRCh38
rs61732591 Chr.19: 52614296-52614296 on Build GRCh38
rs1012784066 Chr.19: 52614359-52614359 on Build GRCh38
rs527276011 Chr.19: 52614218-52614218 on Build GRCh38

View Full Product Details