Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64917353-64917578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKDD1A
Entrez Gene ID
348094
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
-
UniGene
Hs.207157
Gene Chromosome Location
Chr.15: 64911902-64958700 on Build GRCh38
Transcripts
NM_182703.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1301334 c.226G>A p.E76K Chr.15: 64917473-64917473 on Build GRCh38
3887304 c.247C>T p.L83F Chr.15: 64917494-64917494 on Build GRCh38
6464017 c.251C>A p.T84K Chr.15: 64917498-64917498 on Build GRCh38
3936818 c.194G>A p.R65H Chr.15: 64917441-64917441 on Build GRCh38
1374058 c.193C>T p.R65C Chr.15: 64917440-64917440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777675671 Chr.15: 64917440-64917440 on Build GRCh38
rs767731879 Chr.15: 64917507-64917507 on Build GRCh38
rs112321335 Chr.15: 64917405-64917405 on Build GRCh38
rs371733597 Chr.15: 64917377-64917377 on Build GRCh38
rs751753441 Chr.15: 64917413-64917413 on Build GRCh38
rs988692848 Chr.15: 64917400-64917400 on Build GRCh38
rs758990590 Chr.15: 64917402-64917402 on Build GRCh38
rs767047572 Chr.15: 64917491-64917491 on Build GRCh38
rs377565868 Chr.15: 64917483-64917483 on Build GRCh38
rs771575834 Chr.15: 64917462-64917462 on Build GRCh38
rs374847520 Chr.15: 64917441-64917441 on Build GRCh38
rs3057940 Chr.15: 64917520-64917521 on Build GRCh38
rs397837384 Chr.15: 64917540-64917541 on Build GRCh38
rs752499145 Chr.15: 64917520-64917523 on Build GRCh38
rs909777928 Chr.15: 64917536-64917536 on Build GRCh38
rs781358566 Chr.15: 64917427-64917427 on Build GRCh38
rs997883712 Chr.15: 64917519-64917519 on Build GRCh38
rs1043969770 Chr.15: 64917455-64917455 on Build GRCh38
rs558493393 Chr.15: 64917474-64917474 on Build GRCh38
rs566638965 Chr.15: 64917380-64917380 on Build GRCh38
rs545706604 Chr.15: 64917379-64917379 on Build GRCh38
rs953853862 Chr.15: 64917546-64917546 on Build GRCh38
rs369578168 Chr.15: 64917492-64917492 on Build GRCh38
rs770923850 Chr.15: 64917449-64917449 on Build GRCh38
rs1031355281 Chr.15: 64917479-64917479 on Build GRCh38
rs201051822 Chr.15: 64917527-64917527 on Build GRCh38
rs201773834 Chr.15: 64917472-64917472 on Build GRCh38
rs752701846 Chr.15: 64917518-64917518 on Build GRCh38
rs999839839 Chr.15: 64917473-64917473 on Build GRCh38
rs76663839 Chr.15: 64917506-64917506 on Build GRCh38
rs774259704 Chr.15: 64917457-64917457 on Build GRCh38
rs906643562 Chr.15: 64917448-64917448 on Build GRCh38
rs753726615 Chr.15: 64917554-64917554 on Build GRCh38
rs748437776 Chr.15: 64917428-64917428 on Build GRCh38
rs745748638 Chr.15: 64917458-64917458 on Build GRCh38
rs918639869 Chr.15: 64917397-64917397 on Build GRCh38
rs34503100 Chr.15: 64917519-64917520 on Build GRCh38
rs766283052 Chr.15: 64917523-64917524 on Build GRCh38
rs369950609 Chr.15: 64917374-64917374 on Build GRCh38
rs755091969 Chr.15: 64917419-64917419 on Build GRCh38
rs767216330 Chr.15: 64917376-64917376 on Build GRCh38
rs765191352 Chr.15: 64917510-64917511 on Build GRCh38
rs1037010057 Chr.15: 64917382-64917382 on Build GRCh38
rs775286563 Chr.15: 64917484-64917484 on Build GRCh38
rs756245291 Chr.15: 64917437-64917437 on Build GRCh38

View Full Product Details