Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144622993-144623258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPK1
Entrez Gene ID
27010
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
HTPK1, PP20, THMD5
UniGene
Hs.660232
Gene Chromosome Location
Chr.7: 144451941-144836424 on Build GRCh38
Transcripts
NM_001042482.1 NM_022445.3 XM_005249970.1 XM_011516031.1 XM_011516032.2 XM_011516033.2 XM_011516034.2 XM_011516035.2 XM_011516037.2 XM_011516039.2 XM_011516040.2 XM_011516041.2 XM_011516043.1 XM_011516044.1 XM_011516046.1 XM_011516047.2 XM_011516048.1 XM_011516050.1 XM_017011969.1 XM_017011970.1 XM_017011971.1 XM_017011972.1 XM_017011973.1 XM_017011974.1 XM_017011975.1 XM_017011976.1 XM_017011977.1 XM_017011978.1 XM_017011979.1 XM_017011980.1 XM_017011981.1 XM_017011982.1 XM_017011983.1 XM_017011984.1 XM_017011985.1 XM_017011986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3259250 c.353A>C p.K118T Chr.7: 144623167-144623167 on Build GRCh38
745070 c.325C>A p.Q109K Chr.7: 144623195-144623195 on Build GRCh38
4453584 c.317A>T p.K106I Chr.7: 144623203-144623203 on Build GRCh38
421556 c.354+1G>A p.? Chr.7: 144623165-144623165 on Build GRCh38
3635613 c.275C>T p.S92L Chr.7: 144623245-144623245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889867860 Chr.7: 144623033-144623034 on Build GRCh38
rs769684420 Chr.7: 144623100-144623100 on Build GRCh38
rs1014659008 Chr.7: 144623056-144623056 on Build GRCh38
rs968488262 Chr.7: 144623104-144623104 on Build GRCh38
rs767700531 Chr.7: 144623147-144623147 on Build GRCh38
rs1045769598 Chr.7: 144623166-144623166 on Build GRCh38
rs367978008 Chr.7: 144623130-144623130 on Build GRCh38
rs761875562 Chr.7: 144623042-144623042 on Build GRCh38
rs1005884556 Chr.7: 144623045-144623045 on Build GRCh38
rs376729026 Chr.7: 144623043-144623043 on Build GRCh38
rs863224238 Chr.7: 144623209-144623209 on Build GRCh38
rs1000238089 Chr.7: 144623034-144623034 on Build GRCh38
rs755706653 Chr.7: 144623157-144623157 on Build GRCh38
rs779798917 Chr.7: 144623165-144623165 on Build GRCh38
rs778868865 Chr.7: 144623215-144623215 on Build GRCh38
rs748927409 Chr.7: 144623193-144623193 on Build GRCh38
rs888024693 Chr.7: 144623174-144623174 on Build GRCh38
rs1054831762 Chr.7: 144623034-144623034 on Build GRCh38
rs749999689 Chr.7: 144623145-144623145 on Build GRCh38
rs1030137734 Chr.7: 144623066-144623066 on Build GRCh38
rs186564802 Chr.7: 144623194-144623194 on Build GRCh38
rs746011122 Chr.7: 144623227-144623227 on Build GRCh38
rs923899885 Chr.7: 144623120-144623120 on Build GRCh38
rs892052001 Chr.7: 144623052-144623053 on Build GRCh38
rs953186202 Chr.7: 144623086-144623086 on Build GRCh38
rs377118052 Chr.7: 144623125-144623125 on Build GRCh38
rs754758107 Chr.7: 144623214-144623214 on Build GRCh38
rs776838304 Chr.7: 144623224-144623224 on Build GRCh38
rs986481686 Chr.7: 144623097-144623097 on Build GRCh38
rs17170295 Chr.7: 144623121-144623121 on Build GRCh38
rs374277948 Chr.7: 144623114-144623114 on Build GRCh38
rs116240723 Chr.7: 144623118-144623118 on Build GRCh38
rs997181075 Chr.7: 144623065-144623065 on Build GRCh38
rs942240100 Chr.7: 144623225-144623225 on Build GRCh38
rs766944414 Chr.7: 144623129-144623129 on Build GRCh38
rs80241517 Chr.7: 144623063-144623063 on Build GRCh38
rs748032477 Chr.7: 144623220-144623220 on Build GRCh38
rs772204656 Chr.7: 144623223-144623223 on Build GRCh38

View Full Product Details