Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100870551-100870810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIP6
Entrez Gene ID
7205
Gene Name
thyroid hormone receptor interactor 6
Gene Aliases
OIP-1, OIP1, TRIP-6, TRIP6i2, ZRP-1
UniGene
Hs.534360
Gene Chromosome Location
Chr.7: 100867328-100873454 on Build GRCh38
Transcripts
NM_003302.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3876498 c.892C>T p.R298C Chr.7: 100870636-100870636 on Build GRCh38
3876499 c.925A>G p.T309A Chr.7: 100870669-100870669 on Build GRCh38
5453193 c.918_919insG p.C307fs*83 Chr.7: 100870662-100870663 on Build GRCh38
6808910 c.982_983insG p.C328fs*62 Chr.7: 100870726-100870727 on Build GRCh38
5476721 c.830-3C>T p.? Chr.7: 100870571-100870571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552342163 Chr.7: 100870638-100870638 on Build GRCh38
rs768035483 Chr.7: 100870785-100870785 on Build GRCh38
rs546644102 Chr.7: 100870687-100870687 on Build GRCh38
rs775331619 Chr.7: 100870760-100870760 on Build GRCh38
rs779103939 Chr.7: 100870746-100870747 on Build GRCh38
rs367843229 Chr.7: 100870746-100870746 on Build GRCh38
rs767420602 Chr.7: 100870690-100870690 on Build GRCh38
rs536156931 Chr.7: 100870734-100870734 on Build GRCh38
rs751461403 Chr.7: 100870582-100870582 on Build GRCh38
rs760154234 Chr.7: 100870693-100870693 on Build GRCh38
rs755787950 Chr.7: 100870616-100870616 on Build GRCh38
rs760695673 Chr.7: 100870771-100870771 on Build GRCh38
rs781604997 Chr.7: 100870728-100870728 on Build GRCh38
rs749505433 Chr.7: 100870575-100870575 on Build GRCh38
rs1010220171 Chr.7: 100870788-100870788 on Build GRCh38
rs759406705 Chr.7: 100870689-100870689 on Build GRCh38
rs750780514 Chr.7: 100870636-100870636 on Build GRCh38
rs769725224 Chr.7: 100870740-100870740 on Build GRCh38
rs761489431 Chr.7: 100870777-100870777 on Build GRCh38
rs530661397 Chr.7: 100870590-100870590 on Build GRCh38
rs1054391 Chr.7: 100870662-100870662 on Build GRCh38
rs751955583 Chr.7: 100870783-100870783 on Build GRCh38
rs7783543 Chr.7: 100870776-100870776 on Build GRCh38
rs150669875 Chr.7: 100870647-100870647 on Build GRCh38
rs17855370 Chr.7: 100870630-100870630 on Build GRCh38
rs141927859 Chr.7: 100870635-100870635 on Build GRCh38
rs367861588 Chr.7: 100870705-100870705 on Build GRCh38
rs749440517 Chr.7: 100870789-100870789 on Build GRCh38
rs774128661 Chr.7: 100870752-100870752 on Build GRCh38
rs1027087919 Chr.7: 100870745-100870745 on Build GRCh38
rs755534293 Chr.7: 100870716-100870716 on Build GRCh38
rs371080725 Chr.7: 100870675-100870675 on Build GRCh38
rs778782117 Chr.7: 100870637-100870637 on Build GRCh38
rs568026347 Chr.7: 100870707-100870707 on Build GRCh38
rs373669348 Chr.7: 100870742-100870742 on Build GRCh38
rs772024645 Chr.7: 100870759-100870759 on Build GRCh38
rs755439619 Chr.7: 100870784-100870784 on Build GRCh38
rs377078693 Chr.7: 100870639-100870639 on Build GRCh38
rs375530812 Chr.7: 100870648-100870648 on Build GRCh38
rs766960917 Chr.7: 100870591-100870591 on Build GRCh38
rs763772497 Chr.7: 100870697-100870697 on Build GRCh38
rs745620864 Chr.7: 100870786-100870787 on Build GRCh38
rs1034242833 Chr.7: 100870659-100870659 on Build GRCh38
rs985895063 Chr.7: 100870748-100870748 on Build GRCh38
rs953691970 Chr.7: 100870604-100870604 on Build GRCh38
rs148173409 Chr.7: 100870613-100870613 on Build GRCh38
rs753098596 Chr.7: 100870787-100870787 on Build GRCh38
rs372095683 Chr.7: 100870741-100870741 on Build GRCh38
rs763196707 Chr.7: 100870676-100870676 on Build GRCh38
rs961878528 Chr.7: 100870709-100870709 on Build GRCh38
rs759384513 Chr.7: 100870757-100870757 on Build GRCh38
rs773353370 Chr.7: 100870668-100870668 on Build GRCh38
rs754736191 Chr.7: 100870621-100870621 on Build GRCh38
rs748287850 Chr.7: 100870780-100870781 on Build GRCh38
rs765045272 Chr.7: 100870780-100870780 on Build GRCh38
rs917626833 Chr.7: 100870679-100870679 on Build GRCh38
rs755316839 Chr.7: 100870593-100870595 on Build GRCh38
rs375599172 Chr.7: 100870702-100870702 on Build GRCh38
rs375751118 Chr.7: 100870704-100870704 on Build GRCh38
rs752238721 Chr.7: 100870692-100870692 on Build GRCh38
rs776438854 Chr.7: 100870785-100870786 on Build GRCh38

View Full Product Details