Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125998523-125998764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT11
Entrez Gene ID
60487
Gene Name
tRNA methyltransferase 11 homolog
Gene Aliases
C6orf75, MDS024, TRM11, TRMT11-1
UniGene
Hs.404186
Gene Chromosome Location
Chr.6: 125986430-126083131 on Build GRCh38
Transcripts
NM_001031712.2 XM_017011168.1 XM_017011169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470725 c.492G>A p.E164E Chr.6: 125998654-125998654 on Build GRCh38
6470726 c.492G>A p.E164E Chr.6: 125998654-125998654 on Build GRCh38
6808970 c.438A>G p.Q146Q Chr.6: 125998600-125998600 on Build GRCh38
6808969 c.438A>G p.Q146Q Chr.6: 125998600-125998600 on Build GRCh38
1268622 c.442G>C p.V148L Chr.6: 125998604-125998604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772831627 Chr.6: 125998637-125998637 on Build GRCh38
rs147300811 Chr.6: 125998628-125998628 on Build GRCh38
rs780508580 Chr.6: 125998627-125998627 on Build GRCh38
rs758240892 Chr.6: 125998615-125998615 on Build GRCh38
rs746879380 Chr.6: 125998619-125998619 on Build GRCh38
rs145554801 Chr.6: 125998601-125998601 on Build GRCh38
rs373895844 Chr.6: 125998638-125998638 on Build GRCh38
rs571690618 Chr.6: 125998650-125998650 on Build GRCh38
rs546952824 Chr.6: 125998583-125998583 on Build GRCh38
rs761950878 Chr.6: 125998673-125998673 on Build GRCh38
rs772916886 Chr.6: 125998552-125998552 on Build GRCh38
rs758724635 Chr.6: 125998723-125998723 on Build GRCh38
rs34593998 Chr.6: 125998575-125998576 on Build GRCh38
rs192002782 Chr.6: 125998692-125998692 on Build GRCh38
rs186317882 Chr.6: 125998597-125998597 on Build GRCh38
rs764651301 Chr.6: 125998662-125998662 on Build GRCh38
rs772386714 Chr.6: 125998640-125998640 on Build GRCh38
rs766209486 Chr.6: 125998726-125998726 on Build GRCh38
rs765453710 Chr.6: 125998714-125998714 on Build GRCh38
rs759864853 Chr.6: 125998574-125998574 on Build GRCh38
rs750620405 Chr.6: 125998719-125998719 on Build GRCh38
rs879138630 Chr.6: 125998718-125998719 on Build GRCh38
rs370187248 Chr.6: 125998596-125998596 on Build GRCh38
rs780013154 Chr.6: 125998617-125998617 on Build GRCh38
rs750712133 Chr.6: 125998606-125998606 on Build GRCh38
rs775918686 Chr.6: 125998737-125998737 on Build GRCh38
rs6569449 Chr.6: 125998735-125998735 on Build GRCh38
rs753967433 Chr.6: 125998598-125998598 on Build GRCh38
rs754947388 Chr.6: 125998622-125998622 on Build GRCh38
rs1051862121 Chr.6: 125998695-125998695 on Build GRCh38
rs73592112 Chr.6: 125998665-125998665 on Build GRCh38
rs377151003 Chr.6: 125998647-125998647 on Build GRCh38
rs769335142 Chr.6: 125998634-125998634 on Build GRCh38
rs933507254 Chr.6: 125998689-125998689 on Build GRCh38
rs774207339 Chr.6: 125998684-125998684 on Build GRCh38
rs765548651 Chr.6: 125998602-125998602 on Build GRCh38

View Full Product Details