Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41853329-41853592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTBN5
Entrez Gene ID
51332
Gene Name
spectrin beta, non-erythrocytic 5
Gene Aliases
BSPECV, HUBSPECV, HUSPECV
UniGene
Hs.709819
Gene Chromosome Location
Chr.15: 41848146-41894077 on Build GRCh38
Transcripts
NM_016642.3 XM_017022299.1 XM_017022300.1 XM_017022301.1 XM_017022302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4651216 c.10011G>A p.A3337A Chr.15: 41853417-41853417 on Build GRCh38
3719694 c.10018G>A p.E3340K Chr.15: 41853410-41853410 on Build GRCh38
4803038 c.10061G>A p.G3354E Chr.15: 41853367-41853367 on Build GRCh38
117942 c.10098C>A p.C3366* Chr.15: 41853330-41853330 on Build GRCh38
5539700 c.10020G>A p.E3340E Chr.15: 41853408-41853408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751154431 Chr.15: 41853392-41853392 on Build GRCh38
rs748961271 Chr.15: 41853493-41853494 on Build GRCh38
rs71431836 Chr.15: 41853502-41853503 on Build GRCh38
rs777380695 Chr.15: 41853492-41853493 on Build GRCh38
rs778125157 Chr.15: 41853414-41853414 on Build GRCh38
rs999857797 Chr.15: 41853460-41853460 on Build GRCh38
rs746776332 Chr.15: 41853411-41853411 on Build GRCh38
rs904303516 Chr.15: 41853434-41853434 on Build GRCh38
rs568310189 Chr.15: 41853527-41853527 on Build GRCh38
rs187263353 Chr.15: 41853387-41853387 on Build GRCh38
rs776272157 Chr.15: 41853350-41853350 on Build GRCh38
rs149954264 Chr.15: 41853395-41853395 on Build GRCh38
rs774450771 Chr.15: 41853486-41853486 on Build GRCh38
rs756981128 Chr.15: 41853537-41853537 on Build GRCh38
rs74874936 Chr.15: 41853494-41853494 on Build GRCh38
rs546709810 Chr.15: 41853417-41853417 on Build GRCh38
rs371066277 Chr.15: 41853545-41853545 on Build GRCh38
rs987041318 Chr.15: 41853443-41853443 on Build GRCh38
rs1019304848 Chr.15: 41853499-41853499 on Build GRCh38
rs747204416 Chr.15: 41853431-41853431 on Build GRCh38
rs761968364 Chr.15: 41853568-41853568 on Build GRCh38
rs112040796 Chr.15: 41853388-41853388 on Build GRCh38
rs1024210628 Chr.15: 41853349-41853349 on Build GRCh38
rs531292146 Chr.15: 41853432-41853432 on Build GRCh38
rs200316020 Chr.15: 41853410-41853410 on Build GRCh38
rs77259747 Chr.15: 41853464-41853464 on Build GRCh38
rs752338741 Chr.15: 41853496-41853496 on Build GRCh38
rs376144282 Chr.15: 41853393-41853393 on Build GRCh38
rs554848261 Chr.15: 41853511-41853511 on Build GRCh38
rs554221210 Chr.15: 41853533-41853533 on Build GRCh38
rs193289116 Chr.15: 41853480-41853480 on Build GRCh38
rs752143933 Chr.15: 41853400-41853417 on Build GRCh38
rs747246759 Chr.15: 41853571-41853571 on Build GRCh38
rs202132844 Chr.15: 41853401-41853401 on Build GRCh38
rs533462888 Chr.15: 41853551-41853551 on Build GRCh38
rs755689925 Chr.15: 41853433-41853434 on Build GRCh38
rs754243322 Chr.15: 41853549-41853549 on Build GRCh38
rs1004507372 Chr.15: 41853516-41853516 on Build GRCh38
rs772072912 Chr.15: 41853420-41853420 on Build GRCh38
rs566479701 Chr.15: 41853449-41853449 on Build GRCh38
rs761626912 Chr.15: 41853360-41853360 on Build GRCh38
rs534161243 Chr.15: 41853531-41853531 on Build GRCh38
rs568853031 Chr.15: 41853492-41853492 on Build GRCh38
rs373515361 Chr.15: 41853567-41853567 on Build GRCh38
rs764943581 Chr.15: 41853364-41853364 on Build GRCh38
rs757743523 Chr.15: 41853552-41853552 on Build GRCh38
rs866219911 Chr.15: 41853442-41853442 on Build GRCh38
rs779347902 Chr.15: 41853558-41853558 on Build GRCh38
rs773207372 Chr.15: 41853451-41853451 on Build GRCh38
rs975017419 Chr.15: 41853522-41853522 on Build GRCh38
rs754333184 Chr.15: 41853415-41853415 on Build GRCh38
rs750895586 Chr.15: 41853559-41853559 on Build GRCh38
rs975217259 Chr.15: 41853406-41853406 on Build GRCh38
rs565164416 Chr.15: 41853352-41853352 on Build GRCh38
rs201152285 Chr.15: 41853418-41853418 on Build GRCh38
rs780253555 Chr.15: 41853428-41853428 on Build GRCh38

View Full Product Details