Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135449832-135450065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIFAB
Entrez Gene ID
497189
Gene Name
TIFA inhibitor
Gene Aliases
-
UniGene
Hs.552091
Gene Chromosome Location
Chr.5: 135448868-135452399 on Build GRCh38
Transcripts
NM_001099221.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4422499 c.67G>A p.A23T Chr.5: 135449873-135449873 on Build GRCh38
5995873 c.18C>T p.T6T Chr.5: 135449922-135449922 on Build GRCh38
6019043 c.97A>C p.T33P Chr.5: 135449843-135449843 on Build GRCh38
3608633 c.80C>A p.P27Q Chr.5: 135449860-135449860 on Build GRCh38
735641 c.3G>T p.M1I Chr.5: 135449937-135449937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977823800 Chr.5: 135449866-135449866 on Build GRCh38
rs986600849 Chr.5: 135449855-135449855 on Build GRCh38
rs1038610478 Chr.5: 135449922-135449922 on Build GRCh38
rs747546290 Chr.5: 135449852-135449852 on Build GRCh38
rs770787898 Chr.5: 135449978-135449978 on Build GRCh38
rs759895140 Chr.5: 135449963-135449964 on Build GRCh38
rs568164736 Chr.5: 135449923-135449923 on Build GRCh38
rs546746451 Chr.5: 135449914-135449914 on Build GRCh38
rs374967532 Chr.5: 135449854-135449854 on Build GRCh38
rs922362633 Chr.5: 135449883-135449883 on Build GRCh38
rs767281626 Chr.5: 135449892-135449892 on Build GRCh38
rs375528555 Chr.5: 135449932-135449932 on Build GRCh38
rs765885740 Chr.5: 135449904-135449904 on Build GRCh38
rs926686640 Chr.5: 135449912-135449912 on Build GRCh38
rs994158442 Chr.5: 135449990-135449990 on Build GRCh38
rs941597308 Chr.5: 135449916-135449916 on Build GRCh38
rs767795639 Chr.5: 135450005-135450005 on Build GRCh38
rs369279711 Chr.5: 135449857-135449857 on Build GRCh38
rs773210556 Chr.5: 135449967-135449967 on Build GRCh38
rs369737428 Chr.5: 135449934-135449934 on Build GRCh38
rs189922886 Chr.5: 135450030-135450030 on Build GRCh38
rs753222184 Chr.5: 135449966-135449967 on Build GRCh38
rs934193260 Chr.5: 135449885-135449885 on Build GRCh38
rs527797446 Chr.5: 135449893-135449893 on Build GRCh38
rs1003818246 Chr.5: 135450041-135450041 on Build GRCh38
rs372509947 Chr.5: 135449858-135449858 on Build GRCh38
rs749270425 Chr.5: 135449869-135449869 on Build GRCh38
rs779016545 Chr.5: 135449943-135449943 on Build GRCh38
rs760317727 Chr.5: 135449976-135449976 on Build GRCh38
rs764859271 Chr.5: 135449910-135449910 on Build GRCh38
rs907280566 Chr.5: 135450031-135450031 on Build GRCh38
rs768729896 Chr.5: 135449875-135449875 on Build GRCh38
rs754494229 Chr.5: 135449909-135449909 on Build GRCh38
rs560199155 Chr.5: 135449888-135449888 on Build GRCh38
rs200405958 Chr.5: 135449915-135449915 on Build GRCh38
rs550592049 Chr.5: 135450024-135450024 on Build GRCh38
rs768676811 Chr.5: 135449942-135449942 on Build GRCh38
rs769977158 Chr.5: 135449860-135449860 on Build GRCh38
rs920586115 Chr.5: 135449861-135449861 on Build GRCh38
rs529146149 Chr.5: 135449966-135449966 on Build GRCh38
rs753530136 Chr.5: 135449906-135449906 on Build GRCh38
rs775627488 Chr.5: 135449868-135449868 on Build GRCh38
rs748050032 Chr.5: 135449959-135449959 on Build GRCh38
rs760374386 Chr.5: 135449902-135449902 on Build GRCh38
rs756516457 Chr.5: 135449930-135449930 on Build GRCh38
rs774233211 Chr.5: 135449882-135449882 on Build GRCh38
rs35947183 Chr.5: 135449954-135449955 on Build GRCh38
rs1043949453 Chr.5: 135449997-135449997 on Build GRCh38
rs765671856 Chr.5: 135449965-135449966 on Build GRCh38
rs930613777 Chr.5: 135449872-135449872 on Build GRCh38
rs759779390 Chr.5: 135450004-135450004 on Build GRCh38

View Full Product Details