Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114674021-114674280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX3
Entrez Gene ID
6926
Gene Name
T-box 3
Gene Aliases
TBX3-ISO, UMS, XHL
UniGene
Hs.744016
Gene Chromosome Location
Chr.12: 114670254-114684164 on Build GRCh38
Transcripts
NM_005996.3 NM_016569.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4471516 c.1722C>T p.S574S Chr.12: 114674213-114674213 on Build GRCh38
6071321 c.1766C>T p.S589F Chr.12: 114674169-114674169 on Build GRCh38
4471900 c.1747C>T p.Q583* Chr.12: 114674188-114674188 on Build GRCh38
4471496 c.1721C>T p.S574F Chr.12: 114674214-114674214 on Build GRCh38
4530646 c.1715G>A p.G572E Chr.12: 114674220-114674220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201695541 Chr.12: 114674222-114674222 on Build GRCh38
rs988247493 Chr.12: 114674090-114674090 on Build GRCh38
rs372029833 Chr.12: 114674238-114674238 on Build GRCh38
rs780623583 Chr.12: 114674149-114674149 on Build GRCh38
rs768293237 Chr.12: 114674216-114674216 on Build GRCh38
rs886049010 Chr.12: 114674219-114674219 on Build GRCh38
rs548288032 Chr.12: 114674204-114674204 on Build GRCh38
rs540579856 Chr.12: 114674048-114674048 on Build GRCh38
rs375482722 Chr.12: 114674199-114674199 on Build GRCh38
rs745430502 Chr.12: 114674198-114674198 on Build GRCh38
rs747911438 Chr.12: 114674160-114674160 on Build GRCh38
rs767500073 Chr.12: 114674242-114674242 on Build GRCh38
rs757023811 Chr.12: 114674259-114674259 on Build GRCh38
rs775584392 Chr.12: 114674245-114674245 on Build GRCh38
rs760080442 Chr.12: 114674237-114674237 on Build GRCh38
rs894810921 Chr.12: 114674083-114674083 on Build GRCh38
rs973625347 Chr.12: 114674084-114674084 on Build GRCh38
rs756293890 Chr.12: 114674152-114674152 on Build GRCh38
rs952559735 Chr.12: 114674258-114674258 on Build GRCh38
rs528110988 Chr.12: 114674180-114674180 on Build GRCh38
rs1038659583 Chr.12: 114674171-114674171 on Build GRCh38
rs112912347 Chr.12: 114674131-114674131 on Build GRCh38
rs533632144 Chr.12: 114674080-114674080 on Build GRCh38
rs757250778 Chr.12: 114674175-114674175 on Build GRCh38
rs201325654 Chr.12: 114674250-114674250 on Build GRCh38
rs773504740 Chr.12: 114674217-114674217 on Build GRCh38
rs74932638 Chr.12: 114674055-114674055 on Build GRCh38
rs112123944 Chr.12: 114674130-114674130 on Build GRCh38
rs377574870 Chr.12: 114674129-114674129 on Build GRCh38
rs530639044 Chr.12: 114674234-114674234 on Build GRCh38
rs751732977 Chr.12: 114674126-114674126 on Build GRCh38
rs746642104 Chr.12: 114674210-114674210 on Build GRCh38
rs927106325 Chr.12: 114674140-114674140 on Build GRCh38
rs749641288 Chr.12: 114674221-114674221 on Build GRCh38
rs766991967 Chr.12: 114674115-114674115 on Build GRCh38
rs753655242 Chr.12: 114674166-114674166 on Build GRCh38
rs374941856 Chr.12: 114674251-114674251 on Build GRCh38
rs753614292 Chr.12: 114674254-114674254 on Build GRCh38
rs994516201 Chr.12: 114674161-114674161 on Build GRCh38
rs564354358 Chr.12: 114674065-114674065 on Build GRCh38

View Full Product Details