Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21293672-21293943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF708
Entrez Gene ID
7562
Gene Name
zinc finger protein 708
Gene Aliases
KOX8, ZNF15, ZNF15L1
UniGene
Hs.466296
Gene Chromosome Location
Chr.19: 21290388-21329439 on Build GRCh38
Transcripts
NM_001297560.1 NM_001297561.1 NM_021269.2 XM_017027201.1 XM_017027202.1 XM_017027203.1 XM_017027204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993663 c.1167G>T p.E389D Chr.19: 21293799-21293799 on Build GRCh38
1271734 c.1194T>C p.G398G Chr.19: 21293772-21293772 on Build GRCh38
5637233 c.1161T>G p.T387T Chr.19: 21293805-21293805 on Build GRCh38
4280593 c.1072C>A p.H358N Chr.19: 21293894-21293894 on Build GRCh38
5855222 c.1165G>A p.E389K Chr.19: 21293801-21293801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769783384 Chr.19: 21293823-21293823 on Build GRCh38
rs145103647 Chr.19: 21293891-21293891 on Build GRCh38
rs773599352 Chr.19: 21293742-21293742 on Build GRCh38
rs777136793 Chr.19: 21293864-21293864 on Build GRCh38
rs201602198 Chr.19: 21293905-21293905 on Build GRCh38
rs781140869 Chr.19: 21293813-21293813 on Build GRCh38
rs752311320 Chr.19: 21293785-21293785 on Build GRCh38
rs745796175 Chr.19: 21293816-21293816 on Build GRCh38
rs777330306 Chr.19: 21293787-21293787 on Build GRCh38
rs754334293 Chr.19: 21293750-21293750 on Build GRCh38
rs569249843 Chr.19: 21293883-21293883 on Build GRCh38
rs774048698 Chr.19: 21293819-21293820 on Build GRCh38
rs774500506 Chr.19: 21293733-21293733 on Build GRCh38
rs879234692 Chr.19: 21293850-21293850 on Build GRCh38
rs767205608 Chr.19: 21293900-21293900 on Build GRCh38
rs757008302 Chr.19: 21293893-21293893 on Build GRCh38
rs907964123 Chr.19: 21293879-21293879 on Build GRCh38
rs1049087611 Chr.19: 21293730-21293730 on Build GRCh38
rs756014973 Chr.19: 21293903-21293903 on Build GRCh38
rs887749308 Chr.19: 21293737-21293737 on Build GRCh38
rs764851080 Chr.19: 21293770-21293770 on Build GRCh38
rs878922020 Chr.19: 21293845-21293845 on Build GRCh38
rs773329134 Chr.19: 21293835-21293835 on Build GRCh38
rs757021530 Chr.19: 21293795-21293795 on Build GRCh38
rs929361839 Chr.19: 21293720-21293720 on Build GRCh38
rs754022568 Chr.19: 21293847-21293847 on Build GRCh38
rs200621544 Chr.19: 21293830-21293830 on Build GRCh38
rs148160981 Chr.19: 21293747-21293747 on Build GRCh38
rs775104891 Chr.19: 21293873-21293873 on Build GRCh38
rs760104733 Chr.19: 21293866-21293866 on Build GRCh38
rs768752452 Chr.19: 21293725-21293725 on Build GRCh38
rs762012854 Chr.19: 21293739-21293739 on Build GRCh38
rs758118251 Chr.19: 21293786-21293786 on Build GRCh38
rs879166597 Chr.19: 21293907-21293907 on Build GRCh38
rs751343201 Chr.19: 21293793-21293793 on Build GRCh38
rs878997520 Chr.19: 21293844-21293844 on Build GRCh38
rs778963257 Chr.19: 21293828-21293828 on Build GRCh38
rs765908698 Chr.19: 21293869-21293869 on Build GRCh38
rs749337474 Chr.19: 21293717-21293717 on Build GRCh38
rs750286624 Chr.19: 21293901-21293901 on Build GRCh38
rs546241599 Chr.19: 21293815-21293815 on Build GRCh38
rs759052560 Chr.19: 21293752-21293752 on Build GRCh38
rs140097230 Chr.19: 21293842-21293842 on Build GRCh38
rs902028112 Chr.19: 21293852-21293852 on Build GRCh38
rs772264549 Chr.19: 21293834-21293834 on Build GRCh38
rs538144502 Chr.19: 21293887-21293887 on Build GRCh38
rs879110706 Chr.19: 21293856-21293856 on Build GRCh38

View Full Product Details