Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4326731-4327004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAP2
Entrez Gene ID
55620
Gene Name
signal transducing adaptor family member 2
Gene Aliases
BKS
UniGene
Hs.194385
Gene Chromosome Location
Chr.19: 4324043-4338877 on Build GRCh38
Transcripts
NM_001013841.1 NM_017720.2 XM_011528123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5513438 c.809C>T p.A270V Chr.19: 4326962-4326962 on Build GRCh38
997506 c.805G>A p.V269M Chr.19: 4326966-4326966 on Build GRCh38
6293808 c.823G>A p.G275S Chr.19: 4326948-4326948 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1036774008 Chr.19: 4326846-4326846 on Build GRCh38
rs892225816 Chr.19: 4326907-4326907 on Build GRCh38
rs1054040321 Chr.19: 4326793-4326793 on Build GRCh38
rs1028880469 Chr.19: 4326912-4326912 on Build GRCh38
rs201146262 Chr.19: 4326951-4326951 on Build GRCh38
rs963239932 Chr.19: 4326881-4326881 on Build GRCh38
rs906595718 Chr.19: 4326847-4326847 on Build GRCh38
rs768922230 Chr.19: 4326949-4326949 on Build GRCh38
rs563662784 Chr.19: 4326864-4326864 on Build GRCh38
rs1042922164 Chr.19: 4326756-4326756 on Build GRCh38
rs953026619 Chr.19: 4326916-4326916 on Build GRCh38
rs577651376 Chr.19: 4326803-4326803 on Build GRCh38
rs779518193 Chr.19: 4326897-4326897 on Build GRCh38
rs573949653 Chr.19: 4326865-4326865 on Build GRCh38
rs917962703 Chr.19: 4326853-4326853 on Build GRCh38
rs924145988 Chr.19: 4326958-4326958 on Build GRCh38
rs945808902 Chr.19: 4326942-4326942 on Build GRCh38
rs543380947 Chr.19: 4326861-4326861 on Build GRCh38
rs766200781 Chr.19: 4326972-4326972 on Build GRCh38
rs369729010 Chr.19: 4326974-4326974 on Build GRCh38
rs895365159 Chr.19: 4326982-4326982 on Build GRCh38
rs199567037 Chr.19: 4326783-4326784 on Build GRCh38
rs936848775 Chr.19: 4326971-4326971 on Build GRCh38
rs547209443 Chr.19: 4326964-4326964 on Build GRCh38
rs911657333 Chr.19: 4326936-4326936 on Build GRCh38
rs760821330 Chr.19: 4326945-4326945 on Build GRCh38
rs1012269969 Chr.19: 4326939-4326963 on Build GRCh38
rs921164765 Chr.19: 4326770-4326770 on Build GRCh38
rs62129306 Chr.19: 4326901-4326901 on Build GRCh38
rs8109451 Chr.19: 4326784-4326784 on Build GRCh38
rs28448291 Chr.19: 4326903-4326903 on Build GRCh38
rs952544875 Chr.19: 4326775-4326775 on Build GRCh38
rs902668045 Chr.19: 4326778-4326778 on Build GRCh38
rs772141224 Chr.19: 4326914-4326914 on Build GRCh38
rs559738930 Chr.19: 4326924-4326924 on Build GRCh38
rs1025020143 Chr.19: 4326826-4326826 on Build GRCh38
rs895187166 Chr.19: 4326801-4326801 on Build GRCh38
rs936859737 Chr.19: 4326895-4326895 on Build GRCh38
rs1019781737 Chr.19: 4326915-4326915 on Build GRCh38

View Full Product Details