Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57978251-57978524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF606
Entrez Gene ID
80095
Gene Name
zinc finger protein 606
Gene Aliases
ZNF328
UniGene
Hs.654967
Gene Chromosome Location
Chr.19: 57977072-58003346 on Build GRCh38
Transcripts
NM_025027.3 XM_005259276.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6831617 c.2162C>A p.S721* Chr.19: 57978518-57978518 on Build GRCh38
5973211 c.2372T>G p.L791R Chr.19: 57978308-57978308 on Build GRCh38
6852857 c.2300G>T p.S767I Chr.19: 57978380-57978380 on Build GRCh38
173782 c.2288G>A p.R763H Chr.19: 57978392-57978392 on Build GRCh38
4912305 c.2218C>T p.H740Y Chr.19: 57978462-57978462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369798758 Chr.19: 57978283-57978283 on Build GRCh38
rs145119728 Chr.19: 57978438-57978438 on Build GRCh38
rs777068804 Chr.19: 57978448-57978448 on Build GRCh38
rs373899403 Chr.19: 57978366-57978366 on Build GRCh38
rs139469830 Chr.19: 57978334-57978334 on Build GRCh38
rs1022705793 Chr.19: 57978385-57978385 on Build GRCh38
rs778515875 Chr.19: 57978372-57978372 on Build GRCh38
rs775804145 Chr.19: 57978491-57978491 on Build GRCh38
rs989013100 Chr.19: 57978423-57978423 on Build GRCh38
rs141186077 Chr.19: 57978368-57978368 on Build GRCh38
rs1001906662 Chr.19: 57978387-57978387 on Build GRCh38
rs770328470 Chr.19: 57978451-57978451 on Build GRCh38
rs747416589 Chr.19: 57978400-57978400 on Build GRCh38
rs747822561 Chr.19: 57978375-57978375 on Build GRCh38
rs769120687 Chr.19: 57978279-57978280 on Build GRCh38
rs147798149 Chr.19: 57978346-57978346 on Build GRCh38
rs149658738 Chr.19: 57978343-57978343 on Build GRCh38
rs1003026088 Chr.19: 57978401-57978401 on Build GRCh38
rs765550499 Chr.19: 57978284-57978284 on Build GRCh38
rs970096241 Chr.19: 57978378-57978378 on Build GRCh38
rs373070463 Chr.19: 57978392-57978392 on Build GRCh38
rs746211680 Chr.19: 57978450-57978450 on Build GRCh38
rs777270429 Chr.19: 57978393-57978393 on Build GRCh38
rs774787414 Chr.19: 57978284-57978284 on Build GRCh38
rs866144061 Chr.19: 57978455-57978455 on Build GRCh38
rs766288683 Chr.19: 57978365-57978365 on Build GRCh38
rs763099473 Chr.19: 57978304-57978304 on Build GRCh38
rs775894130 Chr.19: 57978292-57978292 on Build GRCh38
rs771456057 Chr.19: 57978425-57978425 on Build GRCh38

View Full Product Details