Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49618844-49619109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPOP
Entrez Gene ID
8405
Gene Name
speckle type BTB/POZ protein
Gene Aliases
BTBD32, TEF2
UniGene
Hs.463382
Gene Chromosome Location
Chr.17: 49598884-49678163 on Build GRCh38
Transcripts
NM_001007226.1 NM_001007227.1 NM_001007228.1 NM_001007229.1 NM_001007230.1 NM_003563.3 XM_005257723.4 XM_005257724.4 XM_017025204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3728329 c.413G>C p.R138P Chr.17: 49619048-49619048 on Build GRCh38
4729697 c.412C>T p.R138C Chr.17: 49619049-49619049 on Build GRCh38
95272 c.399C>G p.F133L Chr.17: 49619062-49619062 on Build GRCh38
1290717 c.388G>A p.D130N Chr.17: 49619073-49619073 on Build GRCh38
5821809 c.457G>T p.D153Y Chr.17: 49619004-49619004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777389504 Chr.17: 49619083-49619083 on Build GRCh38
rs113265648 Chr.17: 49619027-49619027 on Build GRCh38
rs184050028 Chr.17: 49618918-49618918 on Build GRCh38
rs1006096444 Chr.17: 49619057-49619057 on Build GRCh38
rs375093038 Chr.17: 49618984-49618984 on Build GRCh38
rs968974699 Chr.17: 49619049-49619049 on Build GRCh38
rs764345128 Chr.17: 49618958-49618958 on Build GRCh38
rs1041177741 Chr.17: 49618920-49618920 on Build GRCh38
rs189515524 Chr.17: 49618945-49618945 on Build GRCh38
rs1036158089 Chr.17: 49618932-49618932 on Build GRCh38
rs762189786 Chr.17: 49618969-49618969 on Build GRCh38
rs755810590 Chr.17: 49619053-49619053 on Build GRCh38
rs754476215 Chr.17: 49618990-49618990 on Build GRCh38
rs193921065 Chr.17: 49619062-49619062 on Build GRCh38
rs760999631 Chr.17: 49618949-49618949 on Build GRCh38
rs914728063 Chr.17: 49619044-49619044 on Build GRCh38
rs767050824 Chr.17: 49619020-49619020 on Build GRCh38
rs752320964 Chr.17: 49619038-49619038 on Build GRCh38
rs930051780 Chr.17: 49618961-49618961 on Build GRCh38
rs775851222 Chr.17: 49618946-49618946 on Build GRCh38
rs762440331 Chr.17: 49618924-49618924 on Build GRCh38
rs745957392 Chr.17: 49618940-49618940 on Build GRCh38
rs981559838 Chr.17: 49618872-49618872 on Build GRCh38
rs777079703 Chr.17: 49618968-49618968 on Build GRCh38
rs368288966 Chr.17: 49618987-49618987 on Build GRCh38
rs888419251 Chr.17: 49619047-49619047 on Build GRCh38

View Full Product Details