Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84425224-84425459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCLG1
Entrez Gene ID
8802
Gene Name
succinate-CoA ligase alpha subunit
Gene Aliases
GALPHA, MTDPS9, SUCLA1
UniGene
Hs.270428
Gene Chromosome Location
Chr.2: 84423523-84459462 on Build GRCh38
Transcripts
NM_003849.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
477682 c.987T>C p.P329P Chr.2: 84425442-84425442 on Build GRCh38
3364837 c.986C>A p.P329H Chr.2: 84425443-84425443 on Build GRCh38
3001046 c.1014+1G>T p.? Chr.2: 84425414-84425414 on Build GRCh38
3426714 c.1005G>A p.T335T Chr.2: 84425424-84425424 on Build GRCh38
3001047 c.1014G>T p.K338N Chr.2: 84425415-84425415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765965664 Chr.2: 84425425-84425425 on Build GRCh38
rs1034134132 Chr.2: 84425270-84425270 on Build GRCh38
rs1048346458 Chr.2: 84425271-84425271 on Build GRCh38
rs914056512 Chr.2: 84425359-84425360 on Build GRCh38
rs371224605 Chr.2: 84425424-84425424 on Build GRCh38
rs1044675062 Chr.2: 84425351-84425351 on Build GRCh38
rs1003215040 Chr.2: 84425394-84425394 on Build GRCh38
rs958266480 Chr.2: 84425284-84425284 on Build GRCh38
rs201008219 Chr.2: 84425391-84425391 on Build GRCh38
rs751243038 Chr.2: 84425375-84425375 on Build GRCh38
rs767950611 Chr.2: 84425419-84425419 on Build GRCh38
rs774763726 Chr.2: 84425403-84425403 on Build GRCh38
rs368943592 Chr.2: 84425409-84425409 on Build GRCh38
rs1011488179 Chr.2: 84425436-84425436 on Build GRCh38
rs376777509 Chr.2: 84425358-84425358 on Build GRCh38
rs371622046 Chr.2: 84425370-84425370 on Build GRCh38
rs909784004 Chr.2: 84425331-84425331 on Build GRCh38
rs947733812 Chr.2: 84425338-84425338 on Build GRCh38
rs966889026 Chr.2: 84425360-84425360 on Build GRCh38
rs374859665 Chr.2: 84425375-84425375 on Build GRCh38

View Full Product Details